Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151828844-151829115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRA1
Entrez Gene ID
2741
Gene Name
glycine receptor alpha 1
Gene Aliases
HKPX1, STHE
UniGene
Hs.121490
Gene Chromosome Location
Chr.5: 151820798-151924836 on Build GRCh38
Transcripts
NM_000171.3 NM_001146040.1 NM_001292000.1 XM_017009349.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6671707 c.947C>T p.A316V Chr.5: 151829033-151829033 on Build GRCh38
6405106 c.941G>T p.W314L Chr.5: 151829039-151829039 on Build GRCh38
3852949 c.925A>G p.K309E Chr.5: 151829055-151829055 on Build GRCh38
6169917 c.1027C>A p.L343I Chr.5: 151828953-151828953 on Build GRCh38
6169916 c.917C>T p.S306F Chr.5: 151829063-151829063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756370176 Chr.5: 151829076-151829076 on Build GRCh38
rs772149374 Chr.5: 151829052-151829058 on Build GRCh38
rs769499149 Chr.5: 151828999-151828999 on Build GRCh38
rs77451630 Chr.5: 151828958-151828958 on Build GRCh38
rs121918410 Chr.5: 151829060-151829060 on Build GRCh38
rs1049545628 Chr.5: 151828947-151828947 on Build GRCh38
rs369031411 Chr.5: 151829078-151829078 on Build GRCh38
rs374354474 Chr.5: 151828883-151828883 on Build GRCh38
rs377190917 Chr.5: 151828990-151828990 on Build GRCh38
rs281864921 Chr.5: 151829059-151829059 on Build GRCh38
rs761583674 Chr.5: 151828992-151828992 on Build GRCh38
rs939849216 Chr.5: 151828928-151828928 on Build GRCh38
rs78363193 Chr.5: 151828948-151828948 on Build GRCh38
rs373113309 Chr.5: 151829081-151829081 on Build GRCh38
rs776477331 Chr.5: 151828988-151828988 on Build GRCh38
rs751088792 Chr.5: 151829034-151829034 on Build GRCh38
rs200218897 Chr.5: 151828959-151828959 on Build GRCh38
rs867618642 Chr.5: 151829038-151829038 on Build GRCh38
rs772672987 Chr.5: 151829006-151829006 on Build GRCh38
rs762310613 Chr.5: 151828933-151828933 on Build GRCh38
rs199618583 Chr.5: 151828949-151828949 on Build GRCh38
rs75463357 Chr.5: 151828939-151828939 on Build GRCh38
rs150716717 Chr.5: 151829065-151829065 on Build GRCh38
rs760496462 Chr.5: 151828894-151828894 on Build GRCh38
rs907615248 Chr.5: 151829077-151829077 on Build GRCh38
rs749319928 Chr.5: 151828880-151828880 on Build GRCh38
rs768129276 Chr.5: 151829075-151829075 on Build GRCh38
rs780066478 Chr.5: 151828964-151828964 on Build GRCh38
rs748216046 Chr.5: 151828960-151828962 on Build GRCh38
rs763830285 Chr.5: 151828903-151828903 on Build GRCh38
rs746998896 Chr.5: 151828971-151828971 on Build GRCh38
rs970075905 Chr.5: 151829002-151829002 on Build GRCh38
rs147156518 Chr.5: 151828940-151828940 on Build GRCh38
rs281864913 Chr.5: 151828950-151828950 on Build GRCh38
rs892060661 Chr.5: 151828963-151828963 on Build GRCh38
rs148221542 Chr.5: 151828952-151828952 on Build GRCh38
rs765756941 Chr.5: 151829026-151829026 on Build GRCh38
rs776389109 Chr.5: 151828909-151828909 on Build GRCh38
rs550493828 Chr.5: 151829082-151829082 on Build GRCh38
rs766598120 Chr.5: 151828919-151828919 on Build GRCh38
rs201501636 Chr.5: 151829092-151829092 on Build GRCh38
rs762554462 Chr.5: 151829008-151829008 on Build GRCh38
rs775228858 Chr.5: 151828884-151828884 on Build GRCh38
rs938722002 Chr.5: 151828873-151828873 on Build GRCh38
rs773327076 Chr.5: 151829061-151829075 on Build GRCh38
rs370613352 Chr.5: 151828874-151828874 on Build GRCh38
rs750009312 Chr.5: 151828923-151828923 on Build GRCh38
rs764794082 Chr.5: 151828922-151828922 on Build GRCh38
rs139213838 Chr.5: 151828934-151828934 on Build GRCh38
rs761495750 Chr.5: 151828915-151828915 on Build GRCh38
rs1035768184 Chr.5: 151829067-151829067 on Build GRCh38
rs774323435 Chr.5: 151828880-151828881 on Build GRCh38
rs1034889612 Chr.5: 151829014-151829014 on Build GRCh38
rs969350117 Chr.5: 151828977-151828978 on Build GRCh38
rs17112272 Chr.5: 151828987-151828987 on Build GRCh38
rs752071816 Chr.5: 151828935-151828935 on Build GRCh38
rs121918418 Chr.5: 151829009-151829009 on Build GRCh38

View Full Product Details