Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92562219-92562460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM133B
Entrez Gene ID
257415
Gene Name
family with sequence similarity 133 member B
Gene Aliases
-
UniGene
Hs.18564
Gene Chromosome Location
Chr.7: 92560758-92590394 on Build GRCh38
Transcripts
NM_001040057.2 NM_001288584.1 NM_152789.3 NR_109929.1 XM_011516004.2 XM_011516005.2

SNP Detail

SNP ID Chromosome Location
rs909589391 Chr.7: 92562386-92562386 on Build GRCh38
rs1056699842 Chr.7: 92562321-92562321 on Build GRCh38
rs200937544 Chr.7: 92562298-92562298 on Build GRCh38
rs758351783 Chr.7: 92562280-92562280 on Build GRCh38
rs117963956 Chr.7: 92562417-92562417 on Build GRCh38
rs139194447 Chr.7: 92562327-92562327 on Build GRCh38
rs769516575 Chr.7: 92562346-92562349 on Build GRCh38
rs535472118 Chr.7: 92562411-92562411 on Build GRCh38
rs556522408 Chr.7: 92562250-92562250 on Build GRCh38
rs764166713 Chr.7: 92562319-92562321 on Build GRCh38
rs780188303 Chr.7: 92562308-92562308 on Build GRCh38
rs6970992 Chr.7: 92562432-92562432 on Build GRCh38
rs555131384 Chr.7: 92562412-92562412 on Build GRCh38

View Full Product Details