Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124819283-124819542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM175B
Entrez Gene ID
23172
Gene Name
family with sequence similarity 175 member B
Gene Aliases
ABRO1, KIAA0157
UniGene
Hs.280695
Gene Chromosome Location
Chr.10: 124801785-124836670 on Build GRCh38
Transcripts
NM_032182.3 XM_011539554.1 XM_017015986.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
915827 c.259C>T p.R87W Chr.10: 124819442-124819442 on Build GRCh38
2021270 c.212A>T p.Y71F Chr.10: 124819395-124819395 on Build GRCh38
5451065 c.260G>A p.R87Q Chr.10: 124819443-124819443 on Build GRCh38
3435440 c.264A>T p.R88S Chr.10: 124819447-124819447 on Build GRCh38
7425638 c.233A>G p.E78G Chr.10: 124819416-124819416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540521850 Chr.10: 124819330-124819330 on Build GRCh38
rs944096102 Chr.10: 124819414-124819414 on Build GRCh38
rs193100852 Chr.10: 124819464-124819464 on Build GRCh38
rs753759934 Chr.10: 124819421-124819421 on Build GRCh38
rs952744019 Chr.10: 124819367-124819367 on Build GRCh38
rs370584292 Chr.10: 124819487-124819487 on Build GRCh38
rs775946339 Chr.10: 124819473-124819473 on Build GRCh38
rs374553426 Chr.10: 124819396-124819396 on Build GRCh38
rs571531184 Chr.10: 124819428-124819428 on Build GRCh38
rs766371938 Chr.10: 124819401-124819401 on Build GRCh38
rs879222515 Chr.10: 124819329-124819329 on Build GRCh38
rs759548859 Chr.10: 124819495-124819495 on Build GRCh38
rs746954492 Chr.10: 124819448-124819448 on Build GRCh38
rs985219686 Chr.10: 124819389-124819389 on Build GRCh38
rs377610141 Chr.10: 124819443-124819443 on Build GRCh38
rs777266690 Chr.10: 124819444-124819444 on Build GRCh38
rs1035469124 Chr.10: 124819323-124819323 on Build GRCh38
rs1002639620 Chr.10: 124819310-124819310 on Build GRCh38
rs200803584 Chr.10: 124819472-124819472 on Build GRCh38
rs199511821 Chr.10: 124819442-124819442 on Build GRCh38
rs11245376 Chr.10: 124819500-124819500 on Build GRCh38
rs750435703 Chr.10: 124819488-124819488 on Build GRCh38
rs765650397 Chr.10: 124819355-124819355 on Build GRCh38
rs972852515 Chr.10: 124819360-124819360 on Build GRCh38
rs371236649 Chr.10: 124819458-124819458 on Build GRCh38
rs750783193 Chr.10: 124819357-124819357 on Build GRCh38
rs911070599 Chr.10: 124819405-124819405 on Build GRCh38
rs367751038 Chr.10: 124819506-124819506 on Build GRCh38
rs147036264 Chr.10: 124819485-124819485 on Build GRCh38
rs757754050 Chr.10: 124819501-124819501 on Build GRCh38
rs757297862 Chr.10: 124819397-124819397 on Build GRCh38
rs763206188 Chr.10: 124819474-124819474 on Build GRCh38
rs1041095675 Chr.10: 124819477-124819477 on Build GRCh38
rs763270719 Chr.10: 124819373-124819373 on Build GRCh38
rs919844235 Chr.10: 124819363-124819363 on Build GRCh38
rs746554866 Chr.10: 124819454-124819454 on Build GRCh38
rs764418244 Chr.10: 124819480-124819480 on Build GRCh38
rs775654468 Chr.10: 124819359-124819359 on Build GRCh38
rs756495808 Chr.10: 124819366-124819366 on Build GRCh38

View Full Product Details