Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 96203342-96203605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT9
Entrez Gene ID
10690
Gene Name
fucosyltransferase 9
Gene Aliases
Fuc-TIX
UniGene
Hs.49117
Gene Chromosome Location
Chr.6: 96015969-96215612 on Build GRCh38
Transcripts
NM_006581.3 XM_011535383.2 XM_011535385.2 XM_017010188.1 XM_017010190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3630645 c.235C>T p.L79F Chr.6: 96203390-96203390 on Build GRCh38
1082691 c.199C>A p.L67M Chr.6: 96203354-96203354 on Build GRCh38
3630647 c.293C>T p.S98L Chr.6: 96203448-96203448 on Build GRCh38
743800 c.376C>A p.P126T Chr.6: 96203531-96203531 on Build GRCh38
6193346 c.233A>T p.D78V Chr.6: 96203388-96203388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749109692 Chr.6: 96203431-96203431 on Build GRCh38
rs769594295 Chr.6: 96203417-96203417 on Build GRCh38
rs774844654 Chr.6: 96203458-96203458 on Build GRCh38
rs775402689 Chr.6: 96203581-96203581 on Build GRCh38
rs139516318 Chr.6: 96203476-96203476 on Build GRCh38
rs565287641 Chr.6: 96203560-96203560 on Build GRCh38
rs144101900 Chr.6: 96203439-96203439 on Build GRCh38
rs779993303 Chr.6: 96203399-96203399 on Build GRCh38
rs764543353 Chr.6: 96203463-96203463 on Build GRCh38
rs773084092 Chr.6: 96203428-96203428 on Build GRCh38
rs570872346 Chr.6: 96203392-96203392 on Build GRCh38
rs376403164 Chr.6: 96203496-96203496 on Build GRCh38
rs777688941 Chr.6: 96203551-96203551 on Build GRCh38
rs765684352 Chr.6: 96203487-96203487 on Build GRCh38
rs868496669 Chr.6: 96203398-96203398 on Build GRCh38
rs375058208 Chr.6: 96203461-96203461 on Build GRCh38
rs762284154 Chr.6: 96203475-96203475 on Build GRCh38
rs759093263 Chr.6: 96203444-96203444 on Build GRCh38
rs754330739 Chr.6: 96203464-96203464 on Build GRCh38
rs199945143 Chr.6: 96203558-96203558 on Build GRCh38
rs576804723 Chr.6: 96203519-96203519 on Build GRCh38
rs79503863 Chr.6: 96203570-96203570 on Build GRCh38
rs746030301 Chr.6: 96203385-96203385 on Build GRCh38
rs142240115 Chr.6: 96203584-96203584 on Build GRCh38
rs148689797 Chr.6: 96203445-96203445 on Build GRCh38
rs751611875 Chr.6: 96203529-96203529 on Build GRCh38
rs748157492 Chr.6: 96203405-96203405 on Build GRCh38
rs755043633 Chr.6: 96203536-96203536 on Build GRCh38
rs758563441 Chr.6: 96203394-96203394 on Build GRCh38
rs190540423 Chr.6: 96203467-96203467 on Build GRCh38
rs145042649 Chr.6: 96203440-96203440 on Build GRCh38

View Full Product Details