Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 161385697-161385970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSTL5
Entrez Gene ID
56884
Gene Name
follistatin like 5
Gene Aliases
-
UniGene
Hs.32452
Gene Chromosome Location
Chr.4: 161383892-162164034 on Build GRCh38
Transcripts
NM_001128427.2 NM_001128428.2 NM_020116.4 XM_011532126.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6667585 c.2352A>G p.E784E Chr.4: 161385939-161385939 on Build GRCh38
5699893 c.2436A>G p.T812T Chr.4: 161385855-161385855 on Build GRCh38
7260168 c.2517T>A p.N839K Chr.4: 161385774-161385774 on Build GRCh38
6667590 c.2444_2445insA p.D816fs*12 Chr.4: 161385846-161385847 on Build GRCh38
3335001 c.2518A>T p.T840S Chr.4: 161385773-161385773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752510724 Chr.4: 161385869-161385869 on Build GRCh38
rs776965466 Chr.4: 161385892-161385892 on Build GRCh38
rs775338849 Chr.4: 161385921-161385921 on Build GRCh38
rs867713984 Chr.4: 161385839-161385839 on Build GRCh38
rs760530893 Chr.4: 161385926-161385926 on Build GRCh38
rs777240415 Chr.4: 161385789-161385789 on Build GRCh38
rs772552857 Chr.4: 161385728-161385728 on Build GRCh38
rs770280281 Chr.4: 161385803-161385803 on Build GRCh38
rs780653236 Chr.4: 161385879-161385879 on Build GRCh38
rs199897436 Chr.4: 161385783-161385783 on Build GRCh38
rs767400500 Chr.4: 161385868-161385868 on Build GRCh38
rs17040984 Chr.4: 161385908-161385908 on Build GRCh38
rs563187263 Chr.4: 161385820-161385820 on Build GRCh38
rs751443756 Chr.4: 161385768-161385768 on Build GRCh38
rs765214111 Chr.4: 161385864-161385864 on Build GRCh38
rs372559754 Chr.4: 161385748-161385748 on Build GRCh38
rs768939144 Chr.4: 161385840-161385840 on Build GRCh38
rs752384275 Chr.4: 161385784-161385784 on Build GRCh38
rs776031059 Chr.4: 161385821-161385821 on Build GRCh38
rs767455460 Chr.4: 161385920-161385920 on Build GRCh38
rs148257997 Chr.4: 161385757-161385757 on Build GRCh38
rs747386555 Chr.4: 161385883-161385883 on Build GRCh38
rs983110936 Chr.4: 161385739-161385739 on Build GRCh38
rs17040982 Chr.4: 161385848-161385848 on Build GRCh38
rs752011014 Chr.4: 161385819-161385819 on Build GRCh38
rs370089550 Chr.4: 161385738-161385738 on Build GRCh38
rs201875506 Chr.4: 161385871-161385871 on Build GRCh38
rs775904717 Chr.4: 161385734-161385734 on Build GRCh38
rs200389910 Chr.4: 161385876-161385876 on Build GRCh38
rs762023699 Chr.4: 161385861-161385861 on Build GRCh38
rs748842658 Chr.4: 161385791-161385791 on Build GRCh38
rs755793404 Chr.4: 161385788-161385788 on Build GRCh38
rs772557364 Chr.4: 161385871-161385871 on Build GRCh38
rs760950961 Chr.4: 161385737-161385737 on Build GRCh38
rs933767663 Chr.4: 161385860-161385860 on Build GRCh38
rs768902703 Chr.4: 161385885-161385885 on Build GRCh38
rs75149124 Chr.4: 161385872-161385872 on Build GRCh38
rs753977385 Chr.4: 161385744-161385744 on Build GRCh38
rs547566509 Chr.4: 161385927-161385927 on Build GRCh38
rs1039402595 Chr.4: 161385830-161385830 on Build GRCh38
rs762969675 Chr.4: 161385918-161385918 on Build GRCh38
rs761074391 Chr.4: 161385827-161385827 on Build GRCh38
rs909778559 Chr.4: 161385771-161385771 on Build GRCh38
rs779532456 Chr.4: 161385804-161385804 on Build GRCh38
rs746356276 Chr.4: 161385815-161385815 on Build GRCh38
rs570353299 Chr.4: 161385929-161385929 on Build GRCh38
rs754822086 Chr.4: 161385772-161385772 on Build GRCh38
rs759435589 Chr.4: 161385866-161385866 on Build GRCh38
rs111918853 Chr.4: 161385907-161385907 on Build GRCh38

View Full Product Details