Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 106612183-106612408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf57
Entrez Gene ID
55086
Gene Name
chromosome X open reading frame 57
Gene Aliases
-
UniGene
Hs.274267
Gene Chromosome Location
Chr.X: 106611930-106679443 on Build GRCh38
Transcripts
NM_001184782.1 NM_018015.5

SNP Detail

SNP ID Chromosome Location
rs759200579 Chr.X: 106612335-106612335 on Build GRCh38
rs764857453 Chr.X: 106612365-106612365 on Build GRCh38
rs761578433 Chr.X: 106612251-106612251 on Build GRCh38
rs753421133 Chr.X: 106612332-106612332 on Build GRCh38
rs778211678 Chr.X: 106612218-106612218 on Build GRCh38
rs758933008 Chr.X: 106612210-106612210 on Build GRCh38
rs199638136 Chr.X: 106612355-106612355 on Build GRCh38
rs753103211 Chr.X: 106612374-106612374 on Build GRCh38
rs771186847 Chr.X: 106612226-106612226 on Build GRCh38
rs768275039 Chr.X: 106612235-106612235 on Build GRCh38
rs773071044 Chr.X: 106612261-106612261 on Build GRCh38
rs772012236 Chr.X: 106612258-106612258 on Build GRCh38
rs774178715 Chr.X: 106612244-106612244 on Build GRCh38
rs760128227 Chr.X: 106612268-106612268 on Build GRCh38
rs781519814 Chr.X: 106612231-106612231 on Build GRCh38
rs765905243 Chr.X: 106612300-106612300 on Build GRCh38
rs1035070717 Chr.X: 106612293-106612293 on Build GRCh38
rs141536949 Chr.X: 106612266-106612266 on Build GRCh38
rs201142543 Chr.X: 106612232-106612232 on Build GRCh38
rs756257741 Chr.X: 106612349-106612349 on Build GRCh38
rs909164266 Chr.X: 106612202-106612202 on Build GRCh38
rs753088225 Chr.X: 106612209-106612209 on Build GRCh38
rs969190349 Chr.X: 106612252-106612252 on Build GRCh38
rs747057484 Chr.X: 106612220-106612220 on Build GRCh38
rs763459684 Chr.X: 106612299-106612299 on Build GRCh38
rs927179757 Chr.X: 106612238-106612238 on Build GRCh38

View Full Product Details