Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49094231-49094500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHH
Entrez Gene ID
50846
Gene Name
desert hedgehog
Gene Aliases
GDXYM, HHG-3, SRXY7
UniGene
Hs.524382
Gene Chromosome Location
Chr.12: 49086656-49094819 on Build GRCh38
Transcripts
NM_021044.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4631561 c.25C>T p.P9S Chr.12: 49094488-49094488 on Build GRCh38
7297292 c.22C>T p.L8L Chr.12: 49094491-49094491 on Build GRCh38
4647622 c.124C>T p.P42S Chr.12: 49094389-49094389 on Build GRCh38
6643275 c.70G>A p.G24R Chr.12: 49094443-49094443 on Build GRCh38
4398398 c.226G>A p.D76N Chr.12: 49094287-49094287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570786183 Chr.12: 49094300-49094300 on Build GRCh38
rs201628124 Chr.12: 49094286-49094286 on Build GRCh38
rs766441095 Chr.12: 49094413-49094414 on Build GRCh38
rs776969450 Chr.12: 49094461-49094461 on Build GRCh38
rs776863896 Chr.12: 49094362-49094362 on Build GRCh38
rs762023711 Chr.12: 49094282-49094282 on Build GRCh38
rs370468314 Chr.12: 49094279-49094279 on Build GRCh38
rs369889651 Chr.12: 49094462-49094462 on Build GRCh38
rs770758131 Chr.12: 49094256-49094256 on Build GRCh38
rs769474974 Chr.12: 49094335-49094335 on Build GRCh38
rs780967031 Chr.12: 49094426-49094426 on Build GRCh38
rs983920781 Chr.12: 49094334-49094334 on Build GRCh38
rs772319373 Chr.12: 49094354-49094354 on Build GRCh38
rs753295441 Chr.12: 49094387-49094387 on Build GRCh38
rs772856431 Chr.12: 49094392-49094392 on Build GRCh38
rs771221683 Chr.12: 49094359-49094359 on Build GRCh38
rs376829299 Chr.12: 49094339-49094339 on Build GRCh38
rs749218781 Chr.12: 49094340-49094340 on Build GRCh38
rs778014372 Chr.12: 49094442-49094442 on Build GRCh38
rs760988670 Chr.12: 49094429-49094429 on Build GRCh38
rs765506943 Chr.12: 49094378-49094378 on Build GRCh38
rs751248803 Chr.12: 49094324-49094324 on Build GRCh38
rs772268378 Chr.12: 49094284-49094284 on Build GRCh38
rs201464596 Chr.12: 49094321-49094321 on Build GRCh38
rs994912276 Chr.12: 49094434-49094434 on Build GRCh38
rs775788539 Chr.12: 49094470-49094470 on Build GRCh38
rs753139514 Chr.12: 49094319-49094319 on Build GRCh38
rs770229988 Chr.12: 49094463-49094463 on Build GRCh38
rs199552954 Chr.12: 49094281-49094281 on Build GRCh38
rs761002696 Chr.12: 49094357-49094357 on Build GRCh38
rs749269627 Chr.12: 49094438-49094438 on Build GRCh38
rs770909824 Chr.12: 49094451-49094451 on Build GRCh38
rs747016829 Chr.12: 49094450-49094450 on Build GRCh38
rs754916960 Chr.12: 49094439-49094439 on Build GRCh38
rs867169482 Chr.12: 49094349-49094349 on Build GRCh38
rs375841437 Chr.12: 49094266-49094266 on Build GRCh38
rs763200816 Chr.12: 49094368-49094369 on Build GRCh38
rs142269288 Chr.12: 49094325-49094325 on Build GRCh38
rs766883694 Chr.12: 49094290-49094290 on Build GRCh38
rs530705549 Chr.12: 49094315-49094315 on Build GRCh38
rs745612962 Chr.12: 49094329-49094329 on Build GRCh38
rs1007853708 Chr.12: 49094380-49094380 on Build GRCh38
rs756909816 Chr.12: 49094424-49094424 on Build GRCh38
rs754182591 Chr.12: 49094296-49094296 on Build GRCh38
rs138814945 Chr.12: 49094403-49094403 on Build GRCh38
rs1026792446 Chr.12: 49094456-49094456 on Build GRCh38
rs201597166 Chr.12: 49094285-49094285 on Build GRCh38
rs751018684 Chr.12: 49094414-49094414 on Build GRCh38
rs755989004 Chr.12: 49094430-49094430 on Build GRCh38
rs779682957 Chr.12: 49094433-49094433 on Build GRCh38
rs146008219 Chr.12: 49094363-49094363 on Build GRCh38

View Full Product Details