Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75343730-75343997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPS2
Entrez Gene ID
84698
Gene Name
calcyphosine 2
Gene Aliases
UG0636c06
UniGene
Hs.733835
Gene Chromosome Location
Chr.12: 75275979-75390941 on Build GRCh38
Transcripts
NM_001286548.1 NM_001286549.1 XM_005269192.1 XM_011538888.1 XM_011538889.2 XM_011538892.1 XM_017020034.1 XM_017020035.1 XM_017020037.1 XM_017020038.1 XM_017020043.1 XM_017020044.1 XM_017020045.1 NM_001304964.1 NM_152779.3 XM_011538134.2 XM_017019134.1

Gene Symbol
GLIPR1L1
Entrez Gene ID
256710
Gene Name
GLI pathogenesis related 1 like 1
Gene Aliases
ALKN2972, PRO7434
UniGene
Hs.567788
Gene Chromosome Location
Chr.12: 75331834-75370390 on Build GRCh38
Transcripts
NM_001286548.1 NM_001286549.1 XM_005269192.1 XM_011538888.1 XM_011538889.2 XM_011538892.1 XM_017020034.1 XM_017020035.1 XM_017020037.1 XM_017020038.1 XM_017020043.1 XM_017020044.1 XM_017020045.1 NM_001304964.1 NM_152779.3 XM_011538134.2 XM_017019134.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
468901 c.391T>G p.C131G Chr.12: 75343909-75343909 on Build GRCh38
4211248 c.328C>G p.P110A Chr.12: 75343846-75343846 on Build GRCh38
1513139 c.358G>A p.E120K Chr.12: 75343876-75343876 on Build GRCh38
5007051 c.361A>C p.T121P Chr.12: 75343879-75343879 on Build GRCh38
4422812 c.303G>T p.W101C Chr.12: 75343821-75343821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767262317 Chr.12: 75343777-75343777 on Build GRCh38
rs934965239 Chr.12: 75343875-75343875 on Build GRCh38
rs1023742009 Chr.12: 75343965-75343965 on Build GRCh38
rs758198122 Chr.12: 75343805-75343816 on Build GRCh38
rs765401135 Chr.12: 75343790-75343790 on Build GRCh38
rs745313083 Chr.12: 75343918-75343918 on Build GRCh38
rs527762752 Chr.12: 75343939-75343939 on Build GRCh38
rs762272047 Chr.12: 75343962-75343962 on Build GRCh38
rs770923919 Chr.12: 75343844-75343844 on Build GRCh38
rs560648029 Chr.12: 75343774-75343774 on Build GRCh38
rs759111332 Chr.12: 75343770-75343770 on Build GRCh38
rs1049804966 Chr.12: 75343957-75343957 on Build GRCh38
rs768478702 Chr.12: 75343950-75343950 on Build GRCh38
rs374086216 Chr.12: 75343759-75343759 on Build GRCh38
rs766464437 Chr.12: 75343891-75343891 on Build GRCh38
rs773841296 Chr.12: 75343854-75343854 on Build GRCh38
rs762394670 Chr.12: 75343868-75343868 on Build GRCh38
rs765852252 Chr.12: 75343870-75343870 on Build GRCh38
rs56713198 Chr.12: 75343815-75343815 on Build GRCh38
rs777201798 Chr.12: 75343867-75343867 on Build GRCh38
rs764644644 Chr.12: 75343787-75343787 on Build GRCh38
rs190589651 Chr.12: 75343812-75343812 on Build GRCh38
rs764519778 Chr.12: 75343864-75343864 on Build GRCh38
rs757132419 Chr.12: 75343910-75343910 on Build GRCh38
rs752934914 Chr.12: 75343778-75343778 on Build GRCh38
rs150024584 Chr.12: 75343788-75343788 on Build GRCh38
rs749195510 Chr.12: 75343843-75343843 on Build GRCh38
rs1005258219 Chr.12: 75343906-75343906 on Build GRCh38
rs201399852 Chr.12: 75343943-75343943 on Build GRCh38
rs914772843 Chr.12: 75343934-75343934 on Build GRCh38
rs777077332 Chr.12: 75343954-75343954 on Build GRCh38
rs143052259 Chr.12: 75343904-75343904 on Build GRCh38
rs180934864 Chr.12: 75343862-75343862 on Build GRCh38
rs777594867 Chr.12: 75343832-75343832 on Build GRCh38
rs771586842 Chr.12: 75343932-75343932 on Build GRCh38
rs971053536 Chr.12: 75343819-75343819 on Build GRCh38
rs746654617 Chr.12: 75343946-75343946 on Build GRCh38
rs759658957 Chr.12: 75343826-75343826 on Build GRCh38
rs148217982 Chr.12: 75343914-75343914 on Build GRCh38
rs146908424 Chr.12: 75343818-75343818 on Build GRCh38
rs749144230 Chr.12: 75343908-75343908 on Build GRCh38
rs758536764 Chr.12: 75343884-75343884 on Build GRCh38
rs577634037 Chr.12: 75343785-75343785 on Build GRCh38
rs757878744 Chr.12: 75343789-75343789 on Build GRCh38
rs745400747 Chr.12: 75343855-75343855 on Build GRCh38
rs139384889 Chr.12: 75343863-75343863 on Build GRCh38
rs747857750 Chr.12: 75343820-75343820 on Build GRCh38
rs750404476 Chr.12: 75343882-75343882 on Build GRCh38
rs145349592 Chr.12: 75343804-75343804 on Build GRCh38
rs751785233 Chr.12: 75343896-75343896 on Build GRCh38

View Full Product Details