Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6028544-6028773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2AK1
Entrez Gene ID
27102
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 1
Gene Aliases
HCR, HRI
UniGene
Hs.520205
Gene Chromosome Location
Chr.7: 6022247-6059229 on Build GRCh38
Transcripts
NM_001134335.1 NM_014413.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3950672 c.1498G>C p.E500Q Chr.7: 6028647-6028647 on Build GRCh38
5039075 c.1511G>T p.G504V Chr.7: 6028634-6028634 on Build GRCh38
4409336 c.1091C>A p.T364K Chr.7: 6028682-6028682 on Build GRCh38
6448027 c.1091C>T p.T364M Chr.7: 6028682-6028682 on Build GRCh38
5038619 c.1141delT p.S381fs*17 Chr.7: 6028632-6028632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376152082 Chr.7: 6028697-6028697 on Build GRCh38
rs371787803 Chr.7: 6028601-6028601 on Build GRCh38
rs769682382 Chr.7: 6028682-6028682 on Build GRCh38
rs144797860 Chr.7: 6028667-6028667 on Build GRCh38
rs756875216 Chr.7: 6028655-6028655 on Build GRCh38
rs768572436 Chr.7: 6028572-6028572 on Build GRCh38
rs532345337 Chr.7: 6028648-6028648 on Build GRCh38
rs770215808 Chr.7: 6028627-6028627 on Build GRCh38
rs759847990 Chr.7: 6028625-6028625 on Build GRCh38
rs746043505 Chr.7: 6028714-6028714 on Build GRCh38
rs368661636 Chr.7: 6028660-6028660 on Build GRCh38
rs771105947 Chr.7: 6028592-6028592 on Build GRCh38
rs747076833 Chr.7: 6028587-6028587 on Build GRCh38
rs947583310 Chr.7: 6028647-6028647 on Build GRCh38
rs763388999 Chr.7: 6028737-6028737 on Build GRCh38
rs138548924 Chr.7: 6028651-6028651 on Build GRCh38
rs182474233 Chr.7: 6028705-6028705 on Build GRCh38
rs372082679 Chr.7: 6028703-6028703 on Build GRCh38
rs777899865 Chr.7: 6028581-6028581 on Build GRCh38
rs748149087 Chr.7: 6028676-6028676 on Build GRCh38
rs149262605 Chr.7: 6028657-6028657 on Build GRCh38
rs566395713 Chr.7: 6028635-6028635 on Build GRCh38
rs1007954194 Chr.7: 6028662-6028662 on Build GRCh38
rs776153903 Chr.7: 6028629-6028629 on Build GRCh38
rs779885923 Chr.7: 6028681-6028681 on Build GRCh38
rs370810725 Chr.7: 6028689-6028689 on Build GRCh38
rs754913840 Chr.7: 6028690-6028691 on Build GRCh38
rs982629359 Chr.7: 6028731-6028731 on Build GRCh38
rs755878275 Chr.7: 6028677-6028677 on Build GRCh38
rs776063827 Chr.7: 6028727-6028727 on Build GRCh38
rs138441452 Chr.7: 6028685-6028685 on Build GRCh38
rs747079321 Chr.7: 6028702-6028702 on Build GRCh38
rs751164433 Chr.7: 6028656-6028656 on Build GRCh38
rs753685999 Chr.7: 6028595-6028596 on Build GRCh38

View Full Product Details