Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 36598054-36598303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEZ2
Entrez Gene ID
9637
Gene Name
fasciculation and elongation protein zeta 2
Gene Aliases
HUM3CL
UniGene
Hs.258563
Gene Chromosome Location
Chr.2: 36552239-36598190 on Build GRCh38
Transcripts
NM_001042548.1 NM_005102.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6306230 c.22C>A p.Q8K Chr.2: 36598121-36598121 on Build GRCh38
5092437 c.48G>C p.P16P Chr.2: 36598095-36598095 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776908001 Chr.2: 36598110-36598110 on Build GRCh38
rs115873839 Chr.2: 36598276-36598276 on Build GRCh38
rs555617731 Chr.2: 36598187-36598188 on Build GRCh38
rs963003249 Chr.2: 36598178-36598178 on Build GRCh38
rs761979385 Chr.2: 36598113-36598113 on Build GRCh38
rs544100164 Chr.2: 36598277-36598277 on Build GRCh38
rs758534377 Chr.2: 36598251-36598251 on Build GRCh38
rs535304437 Chr.2: 36598182-36598183 on Build GRCh38
rs865979157 Chr.2: 36598189-36598189 on Build GRCh38
rs868072617 Chr.2: 36598216-36598216 on Build GRCh38
rs768959814 Chr.2: 36598097-36598097 on Build GRCh38
rs775210062 Chr.2: 36598176-36598199 on Build GRCh38
rs763026946 Chr.2: 36598179-36598179 on Build GRCh38
rs865891568 Chr.2: 36598131-36598131 on Build GRCh38
rs1025777737 Chr.2: 36598111-36598111 on Build GRCh38
rs776961916 Chr.2: 36598185-36598187 on Build GRCh38
rs980665124 Chr.2: 36598208-36598208 on Build GRCh38
rs11441122 Chr.2: 36598188-36598189 on Build GRCh38
rs921851718 Chr.2: 36598227-36598227 on Build GRCh38
rs987354710 Chr.2: 36598242-36598242 on Build GRCh38
rs867934572 Chr.2: 36598202-36598202 on Build GRCh38
rs773237572 Chr.2: 36598158-36598158 on Build GRCh38
rs761038588 Chr.2: 36598080-36598080 on Build GRCh38
rs564847474 Chr.2: 36598259-36598259 on Build GRCh38
rs539797550 Chr.2: 36598183-36598183 on Build GRCh38
rs750884153 Chr.2: 36598189-36598192 on Build GRCh38
rs951646014 Chr.2: 36598130-36598130 on Build GRCh38
rs974417509 Chr.2: 36598217-36598217 on Build GRCh38
rs954587502 Chr.2: 36598241-36598241 on Build GRCh38
rs945887614 Chr.2: 36598275-36598275 on Build GRCh38
rs771423183 Chr.2: 36598142-36598142 on Build GRCh38
rs769590812 Chr.2: 36598216-36598217 on Build GRCh38
rs760236350 Chr.2: 36598179-36598187 on Build GRCh38
rs765276933 Chr.2: 36598125-36598125 on Build GRCh38
rs1016451948 Chr.2: 36598159-36598159 on Build GRCh38
rs148972737 Chr.2: 36598265-36598265 on Build GRCh38
rs969070782 Chr.2: 36598203-36598203 on Build GRCh38
rs867776053 Chr.2: 36598168-36598168 on Build GRCh38
rs866404033 Chr.2: 36598118-36598118 on Build GRCh38
rs1004614309 Chr.2: 36598141-36598141 on Build GRCh38

View Full Product Details