Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36469443-36469702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF3R
Entrez Gene ID
1441
Gene Name
colony stimulating factor 3 receptor
Gene Aliases
CD114, GCSFR, SCN7
UniGene
Hs.524517
Gene Chromosome Location
Chr.1: 36466043-36483314 on Build GRCh38
Transcripts
NM_000760.3 NM_156039.3 NM_172313.2 XM_005270493.1 XM_011540748.2 XM_011540749.1 XM_011540750.1 XM_017000370.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6932459 c.1431G>T p.W477C Chr.1: 36469695-36469695 on Build GRCh38
6932458 c.1431G>T p.W477C Chr.1: 36469695-36469695 on Build GRCh38
1249173 c.1459G>T p.G487W Chr.1: 36469667-36469667 on Build GRCh38
680885 c.1427C>T p.T476I Chr.1: 36469699-36469699 on Build GRCh38
42643 c.1438G>A p.E480K Chr.1: 36469688-36469688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776942736 Chr.1: 36469579-36469579 on Build GRCh38
rs954548866 Chr.1: 36469653-36469653 on Build GRCh38
rs991350060 Chr.1: 36469548-36469548 on Build GRCh38
rs762641978 Chr.1: 36469664-36469664 on Build GRCh38
rs780350557 Chr.1: 36469473-36469473 on Build GRCh38
rs1046488175 Chr.1: 36469512-36469512 on Build GRCh38
rs371482099 Chr.1: 36469648-36469648 on Build GRCh38
rs879757617 Chr.1: 36469504-36469504 on Build GRCh38
rs536895011 Chr.1: 36469472-36469472 on Build GRCh38
rs754914504 Chr.1: 36469601-36469601 on Build GRCh38
rs574358253 Chr.1: 36469526-36469526 on Build GRCh38
rs755985922 Chr.1: 36469615-36469615 on Build GRCh38
rs544773947 Chr.1: 36469674-36469674 on Build GRCh38
rs948135861 Chr.1: 36469470-36469470 on Build GRCh38
rs759506026 Chr.1: 36469669-36469669 on Build GRCh38
rs767341968 Chr.1: 36469603-36469603 on Build GRCh38
rs770239807 Chr.1: 36469606-36469606 on Build GRCh38
rs780048776 Chr.1: 36469632-36469632 on Build GRCh38
rs1011687434 Chr.1: 36469555-36469555 on Build GRCh38
rs544605647 Chr.1: 36469545-36469545 on Build GRCh38
rs772801416 Chr.1: 36469651-36469651 on Build GRCh38
rs767301627 Chr.1: 36469670-36469670 on Build GRCh38
rs199870771 Chr.1: 36469645-36469645 on Build GRCh38
rs1021272735 Chr.1: 36469562-36469562 on Build GRCh38
rs770442353 Chr.1: 36469665-36469665 on Build GRCh38
rs3917989 Chr.1: 36469527-36469527 on Build GRCh38
rs138289951 Chr.1: 36469668-36469668 on Build GRCh38
rs769098766 Chr.1: 36469637-36469637 on Build GRCh38
rs746127142 Chr.1: 36469624-36469624 on Build GRCh38
rs748440449 Chr.1: 36469646-36469646 on Build GRCh38
rs909700624 Chr.1: 36469503-36469503 on Build GRCh38
rs376850709 Chr.1: 36469619-36469619 on Build GRCh38
rs758458810 Chr.1: 36469626-36469626 on Build GRCh38
rs999653811 Chr.1: 36469531-36469531 on Build GRCh38
rs1053157003 Chr.1: 36469539-36469539 on Build GRCh38
rs530239898 Chr.1: 36469586-36469586 on Build GRCh38
rs748269112 Chr.1: 36469581-36469581 on Build GRCh38
rs747544404 Chr.1: 36469633-36469633 on Build GRCh38

View Full Product Details