Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159782118-159782365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP23
Entrez Gene ID
54935
Gene Name
dual specificity phosphatase 23
Gene Aliases
DUSP25, LDP-3, MOSP, VHZ
UniGene
Hs.425801
Gene Chromosome Location
Chr.1: 159780946-159782546 on Build GRCh38
Transcripts
NM_001319658.1 NM_001319659.1 NM_017823.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
24283 c.356G>A p.G119E Chr.1: 159782241-159782241 on Build GRCh38
2084698 c.301C>T p.R101C Chr.1: 159782186-159782186 on Build GRCh38
6508017 c.447G>A p.T149T Chr.1: 159782332-159782332 on Build GRCh38
4167011 c.373A>G p.I125V Chr.1: 159782258-159782258 on Build GRCh38
4491217 c.376C>G p.R126G Chr.1: 159782261-159782261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138366687 Chr.1: 159782262-159782262 on Build GRCh38
rs576265969 Chr.1: 159782142-159782142 on Build GRCh38
rs370697804 Chr.1: 159782270-159782270 on Build GRCh38
rs747457096 Chr.1: 159782191-159782191 on Build GRCh38
rs201443958 Chr.1: 159782148-159782148 on Build GRCh38
rs200214393 Chr.1: 159782225-159782225 on Build GRCh38
rs771307415 Chr.1: 159782292-159782292 on Build GRCh38
rs199923945 Chr.1: 159782136-159782136 on Build GRCh38
rs762808262 Chr.1: 159782144-159782144 on Build GRCh38
rs763777923 Chr.1: 159782298-159782298 on Build GRCh38
rs767381391 Chr.1: 159782254-159782254 on Build GRCh38
rs775288918 Chr.1: 159782317-159782317 on Build GRCh38
rs762029754 Chr.1: 159782138-159782138 on Build GRCh38
rs11544443 Chr.1: 159782256-159782256 on Build GRCh38
rs780331860 Chr.1: 159782265-159782265 on Build GRCh38
rs144974677 Chr.1: 159782170-159782170 on Build GRCh38
rs963369910 Chr.1: 159782249-159782249 on Build GRCh38
rs748756501 Chr.1: 159782271-159782271 on Build GRCh38
rs369974155 Chr.1: 159782162-159782162 on Build GRCh38
rs748982089 Chr.1: 159782172-159782172 on Build GRCh38
rs1046316617 Chr.1: 159782244-159782244 on Build GRCh38
rs756864682 Chr.1: 159782328-159782328 on Build GRCh38
rs773434991 Chr.1: 159782279-159782279 on Build GRCh38
rs1129923 Chr.1: 159782276-159782276 on Build GRCh38
rs998327504 Chr.1: 159782160-159782160 on Build GRCh38
rs760726840 Chr.1: 159782320-159782320 on Build GRCh38
rs529005069 Chr.1: 159782206-159782206 on Build GRCh38
rs777273088 Chr.1: 159782171-159782171 on Build GRCh38
rs989587241 Chr.1: 159782219-159782219 on Build GRCh38
rs11544445 Chr.1: 159782212-159782212 on Build GRCh38
rs747407382 Chr.1: 159782266-159782266 on Build GRCh38
rs759503549 Chr.1: 159782247-159782247 on Build GRCh38
rs192795373 Chr.1: 159782186-159782186 on Build GRCh38
rs538013186 Chr.1: 159782322-159782322 on Build GRCh38
rs147728803 Chr.1: 159782169-159782169 on Build GRCh38
rs780682835 Chr.1: 159782166-159782166 on Build GRCh38
rs926446053 Chr.1: 159782318-159782318 on Build GRCh38
rs977824746 Chr.1: 159782185-159782185 on Build GRCh38
rs759386709 Chr.1: 159782300-159782300 on Build GRCh38
rs559123240 Chr.1: 159782237-159782237 on Build GRCh38
rs763731199 Chr.1: 159782261-159782261 on Build GRCh38
rs200750638 Chr.1: 159782226-159782226 on Build GRCh38
rs774289461 Chr.1: 159782230-159782230 on Build GRCh38
rs552406687 Chr.1: 159782269-159782269 on Build GRCh38
rs756917558 Chr.1: 159782264-159782264 on Build GRCh38
rs368989027 Chr.1: 159782143-159782143 on Build GRCh38
rs755551993 Chr.1: 159782258-159782258 on Build GRCh38
rs753351367 Chr.1: 159782327-159782327 on Build GRCh38
rs867323325 Chr.1: 159782301-159782301 on Build GRCh38
rs937893919 Chr.1: 159782326-159782326 on Build GRCh38
rs749450188 Chr.1: 159782282-159782282 on Build GRCh38
rs748152682 Chr.1: 159782213-159782213 on Build GRCh38
rs763321584 Chr.1: 159782229-159782229 on Build GRCh38
rs764846450 Chr.1: 159782343-159782343 on Build GRCh38
rs751526207 Chr.1: 159782149-159782149 on Build GRCh38
rs774236038 Chr.1: 159782298-159782298 on Build GRCh38
rs758760967 Chr.1: 159782173-159782173 on Build GRCh38

View Full Product Details