Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184572834-184573065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB3
Entrez Gene ID
2049
Gene Name
EPH receptor B3
Gene Aliases
EK2, ETK2, HEK2, TYRO6
UniGene
Hs.2913
Gene Chromosome Location
Chr.3: 184561799-184582408 on Build GRCh38
Transcripts
NM_004443.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5476445 c.517C>T p.R173C Chr.3: 184572837-184572837 on Build GRCh38
4425663 c.617G>A p.R206H Chr.3: 184572937-184572937 on Build GRCh38
1743275 c.658G>A p.A220T Chr.3: 184572978-184572978 on Build GRCh38
3590809 c.571C>T p.L191L Chr.3: 184572891-184572891 on Build GRCh38
7307611 c.716_717insC p.T240fs*5 Chr.3: 184573036-184573037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747329213 Chr.3: 184573007-184573007 on Build GRCh38
rs769299714 Chr.3: 184573019-184573019 on Build GRCh38
rs970108083 Chr.3: 184572952-184572952 on Build GRCh38
rs758025687 Chr.3: 184572939-184572939 on Build GRCh38
rs373196824 Chr.3: 184572968-184572968 on Build GRCh38
rs762220469 Chr.3: 184573039-184573039 on Build GRCh38
rs187877110 Chr.3: 184572969-184572969 on Build GRCh38
rs753310936 Chr.3: 184572932-184572932 on Build GRCh38
rs764652577 Chr.3: 184572924-184572924 on Build GRCh38
rs530187712 Chr.3: 184573022-184573022 on Build GRCh38
rs546016485 Chr.3: 184572923-184572923 on Build GRCh38
rs577684335 Chr.3: 184572986-184572986 on Build GRCh38
rs1026987955 Chr.3: 184572911-184572911 on Build GRCh38
rs772620531 Chr.3: 184572892-184572892 on Build GRCh38
rs112247281 Chr.3: 184572938-184572938 on Build GRCh38
rs746148321 Chr.3: 184572989-184572989 on Build GRCh38
rs982875786 Chr.3: 184572941-184572941 on Build GRCh38
rs757427786 Chr.3: 184572987-184572987 on Build GRCh38
rs759675909 Chr.3: 184572879-184572879 on Build GRCh38
rs140016968 Chr.3: 184572876-184572876 on Build GRCh38
rs772389628 Chr.3: 184572990-184572990 on Build GRCh38
rs145290731 Chr.3: 184572977-184572977 on Build GRCh38
rs779865356 Chr.3: 184572966-184572966 on Build GRCh38
rs371321162 Chr.3: 184572912-184572912 on Build GRCh38
rs776024993 Chr.3: 184572997-184572997 on Build GRCh38
rs376915139 Chr.3: 184572855-184572855 on Build GRCh38
rs1053717016 Chr.3: 184572981-184572981 on Build GRCh38
rs267599717 Chr.3: 184572933-184572933 on Build GRCh38

View Full Product Details