Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 23693182-23693455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD2B
Entrez Gene ID
54454
Gene Name
ATPase family, AAA domain containing 2B
Gene Aliases
-
UniGene
Hs.467862
Gene Chromosome Location
Chr.2: 23689710-23927114 on Build GRCh38
Transcripts
XM_006712030.3 NM_052920.1 XM_006711929.3 XM_011532501.2 XM_017003264.1

Gene Symbol
KLHL29
Entrez Gene ID
114818
Gene Name
kelch like family member 29
Gene Aliases
KBTBD9
UniGene
Hs.130593
Gene Chromosome Location
Chr.2: 23385419-23708613 on Build GRCh38
Transcripts
XM_006712030.3 NM_052920.1 XM_006711929.3 XM_011532501.2 XM_017003264.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1406553 c.1440C>A p.F480L Chr.2: 23693426-23693426 on Build GRCh38
6038281 c.1329C>T p.A443A Chr.2: 23693315-23693315 on Build GRCh38
401935 c.687C>A p.S229R Chr.2: 23693333-23693333 on Build GRCh38
3380100 c.1393G>A p.E465K Chr.2: 23693379-23693379 on Build GRCh38
2908921 c.1330G>A p.E444K Chr.2: 23693316-23693316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34854124 Chr.2: 23693270-23693271 on Build GRCh38
rs1007446750 Chr.2: 23693337-23693337 on Build GRCh38
rs543437540 Chr.2: 23693379-23693379 on Build GRCh38
rs377729877 Chr.2: 23693435-23693435 on Build GRCh38
rs369774606 Chr.2: 23693388-23693388 on Build GRCh38
rs866327882 Chr.2: 23693316-23693316 on Build GRCh38
rs779007488 Chr.2: 23693261-23693261 on Build GRCh38
rs911194368 Chr.2: 23693436-23693436 on Build GRCh38
rs188106214 Chr.2: 23693260-23693260 on Build GRCh38
rs774746015 Chr.2: 23693387-23693387 on Build GRCh38
rs750429982 Chr.2: 23693282-23693282 on Build GRCh38
rs758437509 Chr.2: 23693301-23693301 on Build GRCh38
rs372942747 Chr.2: 23693333-23693333 on Build GRCh38
rs970977648 Chr.2: 23693414-23693414 on Build GRCh38
rs199970151 Chr.2: 23693429-23693429 on Build GRCh38
rs1000662622 Chr.2: 23693375-23693375 on Build GRCh38
rs973302968 Chr.2: 23693418-23693418 on Build GRCh38
rs533521165 Chr.2: 23693253-23693253 on Build GRCh38
rs183179970 Chr.2: 23693231-23693231 on Build GRCh38
rs555352685 Chr.2: 23693254-23693254 on Build GRCh38
rs577898730 Chr.2: 23693360-23693360 on Build GRCh38
rs957288288 Chr.2: 23693370-23693370 on Build GRCh38
rs1038980975 Chr.2: 23693343-23693343 on Build GRCh38
rs374010012 Chr.2: 23693430-23693430 on Build GRCh38
rs955110915 Chr.2: 23693409-23693409 on Build GRCh38
rs573807664 Chr.2: 23693255-23693255 on Build GRCh38
rs78203243 Chr.2: 23693345-23693345 on Build GRCh38
rs756193229 Chr.2: 23693352-23693352 on Build GRCh38
rs191598480 Chr.2: 23693363-23693363 on Build GRCh38
rs918653142 Chr.2: 23693274-23693274 on Build GRCh38
rs115363888 Chr.2: 23693378-23693378 on Build GRCh38
rs778433191 Chr.2: 23693351-23693351 on Build GRCh38
rs979495846 Chr.2: 23693230-23693230 on Build GRCh38
rs931448202 Chr.2: 23693315-23693315 on Build GRCh38
rs918800744 Chr.2: 23693426-23693426 on Build GRCh38

View Full Product Details