Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 100535776-100536035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM174A
Entrez Gene ID
345757
Gene Name
family with sequence similarity 174 member A
Gene Aliases
HGS_RE408, TMEM157, UNQ1912
UniGene
Hs.356108
Gene Chromosome Location
Chr.5: 100535305-100586741 on Build GRCh38
Transcripts
NM_198507.2 XM_006714600.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3857065 c.263T>C p.V88A Chr.5: 100535793-100535793 on Build GRCh38
1193316 c.283G>T p.D95Y Chr.5: 100535813-100535813 on Build GRCh38
5869014 c.304G>A p.E102K Chr.5: 100535834-100535834 on Build GRCh38
738844 c.306A>T p.E102D Chr.5: 100535836-100535836 on Build GRCh38
7313446 c.403G>T p.V135L Chr.5: 100535933-100535933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372498908 Chr.5: 100536000-100536000 on Build GRCh38
rs531483245 Chr.5: 100535818-100535818 on Build GRCh38
rs759880540 Chr.5: 100535947-100535947 on Build GRCh38
rs770904098 Chr.5: 100535874-100535874 on Build GRCh38
rs991987171 Chr.5: 100535942-100535942 on Build GRCh38
rs746111753 Chr.5: 100536002-100536002 on Build GRCh38
rs763670180 Chr.5: 100535826-100535826 on Build GRCh38
rs776908556 Chr.5: 100535959-100535959 on Build GRCh38
rs750071967 Chr.5: 100535977-100535977 on Build GRCh38
rs200948075 Chr.5: 100535980-100535980 on Build GRCh38
rs369516601 Chr.5: 100535993-100535993 on Build GRCh38
rs776960906 Chr.5: 100535937-100535937 on Build GRCh38
rs751002251 Chr.5: 100535846-100535846 on Build GRCh38
rs749237204 Chr.5: 100535894-100535894 on Build GRCh38
rs960008278 Chr.5: 100535914-100535914 on Build GRCh38
rs747035012 Chr.5: 100535930-100535930 on Build GRCh38
rs766666353 Chr.5: 100535797-100535797 on Build GRCh38
rs745716053 Chr.5: 100535870-100535870 on Build GRCh38
rs771129761 Chr.5: 100535996-100535996 on Build GRCh38
rs755921624 Chr.5: 100535981-100535981 on Build GRCh38
rs767202041 Chr.5: 100535973-100535973 on Build GRCh38
rs756741908 Chr.5: 100535964-100535964 on Build GRCh38
rs763518688 Chr.5: 100535951-100535951 on Build GRCh38
rs372797030 Chr.5: 100535900-100535900 on Build GRCh38
rs760994288 Chr.5: 100535794-100535794 on Build GRCh38
rs753238312 Chr.5: 100535820-100535820 on Build GRCh38
rs764547900 Chr.5: 100535958-100535958 on Build GRCh38
rs775952388 Chr.5: 100536015-100536015 on Build GRCh38
rs1011149361 Chr.5: 100535838-100535838 on Build GRCh38
rs566008413 Chr.5: 100535989-100535989 on Build GRCh38
rs115404031 Chr.5: 100536008-100536008 on Build GRCh38
rs756936364 Chr.5: 100535855-100535855 on Build GRCh38
rs751095320 Chr.5: 100535961-100535961 on Build GRCh38
rs372056361 Chr.5: 100535824-100535824 on Build GRCh38
rs375418043 Chr.5: 100535885-100535885 on Build GRCh38
rs754275569 Chr.5: 100535810-100535810 on Build GRCh38
rs755911105 Chr.5: 100535871-100535871 on Build GRCh38
rs765617668 Chr.5: 100535948-100535948 on Build GRCh38
rs796598051 Chr.5: 100535849-100535849 on Build GRCh38
rs774173259 Chr.5: 100536009-100536009 on Build GRCh38
rs780777327 Chr.5: 100535859-100535859 on Build GRCh38
rs201541686 Chr.5: 100535895-100535895 on Build GRCh38
rs139597477 Chr.5: 100535814-100535814 on Build GRCh38
rs773432657 Chr.5: 100535897-100535897 on Build GRCh38
rs969694870 Chr.5: 100535872-100535872 on Build GRCh38
rs771326074 Chr.5: 100535936-100535936 on Build GRCh38
rs185049199 Chr.5: 100535992-100535992 on Build GRCh38

View Full Product Details