Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24947544-24947817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC27
Entrez Gene ID
51277
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Gene Aliases
RBJ, RabJS
UniGene
Hs.434993
Gene Chromosome Location
Chr.2: 24943636-24972094 on Build GRCh38
Transcripts
NM_001198559.1 NM_016544.2 XM_017004269.1 XM_017004270.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5973720 c.779C>A p.A260E Chr.2: 24947659-24947659 on Build GRCh38
4442469 c.770C>T p.A257V Chr.2: 24947668-24947668 on Build GRCh38
4852067 c.766G>A p.D256N Chr.2: 24947672-24947672 on Build GRCh38
5862259 c.758G>A p.G253D Chr.2: 24947680-24947680 on Build GRCh38
253710 c.701A>G p.N234S Chr.2: 24947737-24947737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746918288 Chr.2: 24947644-24947644 on Build GRCh38
rs369759934 Chr.2: 24947751-24947751 on Build GRCh38
rs1015039576 Chr.2: 24947693-24947693 on Build GRCh38
rs547339830 Chr.2: 24947795-24947795 on Build GRCh38
rs978131656 Chr.2: 24947641-24947641 on Build GRCh38
rs759362910 Chr.2: 24947779-24947780 on Build GRCh38
rs756869075 Chr.2: 24947587-24947587 on Build GRCh38
rs758169335 Chr.2: 24947775-24947775 on Build GRCh38
rs200057489 Chr.2: 24947759-24947759 on Build GRCh38
rs750070168 Chr.2: 24947771-24947771 on Build GRCh38
rs774553651 Chr.2: 24947674-24947674 on Build GRCh38
rs201340951 Chr.2: 24947721-24947721 on Build GRCh38
rs764471180 Chr.2: 24947580-24947580 on Build GRCh38
rs768496017 Chr.2: 24947645-24947645 on Build GRCh38
rs754187199 Chr.2: 24947586-24947586 on Build GRCh38
rs1008569629 Chr.2: 24947689-24947689 on Build GRCh38
rs745572390 Chr.2: 24947599-24947599 on Build GRCh38
rs374812991 Chr.2: 24947787-24947787 on Build GRCh38
rs1046332759 Chr.2: 24947756-24947756 on Build GRCh38
rs991339608 Chr.2: 24947777-24947777 on Build GRCh38
rs759151161 Chr.2: 24947622-24947622 on Build GRCh38
rs1029754206 Chr.2: 24947616-24947616 on Build GRCh38
rs765762166 Chr.2: 24947765-24947765 on Build GRCh38
rs375430905 Chr.2: 24947730-24947730 on Build GRCh38
rs776444237 Chr.2: 24947650-24947650 on Build GRCh38
rs779336428 Chr.2: 24947593-24947593 on Build GRCh38
rs761204268 Chr.2: 24947726-24947726 on Build GRCh38
rs565382768 Chr.2: 24947789-24947789 on Build GRCh38
rs149877713 Chr.2: 24947731-24947731 on Build GRCh38
rs2241837 Chr.2: 24947649-24947649 on Build GRCh38
rs756662962 Chr.2: 24947571-24947571 on Build GRCh38
rs368660593 Chr.2: 24947659-24947659 on Build GRCh38
rs772121404 Chr.2: 24947709-24947709 on Build GRCh38
rs766960530 Chr.2: 24947694-24947694 on Build GRCh38
rs748227013 Chr.2: 24947606-24947606 on Build GRCh38
rs376043877 Chr.2: 24947784-24947784 on Build GRCh38
rs779657408 Chr.2: 24947783-24947783 on Build GRCh38
rs779749473 Chr.2: 24947629-24947629 on Build GRCh38

View Full Product Details