Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14743223-14743484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE2
Entrez Gene ID
30817
Gene Name
adhesion G protein-coupled receptor E2
Gene Aliases
CD312, EMR2, VBU
UniGene
Hs.531619
Gene Chromosome Location
Chr.19: 14732697-14778541 on Build GRCh38
Transcripts
NM_001271052.1 NM_013447.3 XM_011527948.2 XM_011527949.2 XM_011527951.2 XM_011527952.2 XM_011527953.2 XM_011527954.2 XM_011527955.2 XM_017026726.1 XM_017026727.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5854822 c.2435C>T p.A812V Chr.19: 14743448-14743448 on Build GRCh38
5022008 c.2462C>T p.T821M Chr.19: 14743421-14743421 on Build GRCh38
3282417 c.2410G>C p.E804Q Chr.19: 14743473-14743473 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs897064648 Chr.19: 14743296-14743296 on Build GRCh38
rs61732002 Chr.19: 14743421-14743421 on Build GRCh38
rs141741521 Chr.19: 14743419-14743419 on Build GRCh38
rs763943686 Chr.19: 14743382-14743382 on Build GRCh38
rs1008474954 Chr.19: 14743268-14743268 on Build GRCh38
rs757200390 Chr.19: 14743394-14743394 on Build GRCh38
rs750461791 Chr.19: 14743409-14743409 on Build GRCh38
rs910243776 Chr.19: 14743284-14743284 on Build GRCh38
rs752646895 Chr.19: 14743278-14743278 on Build GRCh38
rs758293688 Chr.19: 14743380-14743380 on Build GRCh38
rs936937358 Chr.19: 14743295-14743295 on Build GRCh38
rs969656957 Chr.19: 14743358-14743358 on Build GRCh38
rs1036890955 Chr.19: 14743274-14743274 on Build GRCh38
rs781385716 Chr.19: 14743397-14743397 on Build GRCh38
rs557195074 Chr.19: 14743309-14743309 on Build GRCh38
rs906602425 Chr.19: 14743333-14743333 on Build GRCh38
rs575490119 Chr.19: 14743308-14743308 on Build GRCh38
rs1026878246 Chr.19: 14743343-14743343 on Build GRCh38
rs758950184 Chr.19: 14743446-14743446 on Build GRCh38
rs922194314 Chr.19: 14743460-14743460 on Build GRCh38
rs945540969 Chr.19: 14743300-14743300 on Build GRCh38
rs762799719 Chr.19: 14743342-14743342 on Build GRCh38
rs752262553 Chr.19: 14743407-14743408 on Build GRCh38
rs138856853 Chr.19: 14743438-14743438 on Build GRCh38
rs776246646 Chr.19: 14743445-14743445 on Build GRCh38
rs371031119 Chr.19: 14743383-14743383 on Build GRCh38
rs752593078 Chr.19: 14743376-14743376 on Build GRCh38
rs375661729 Chr.19: 14743410-14743410 on Build GRCh38
rs890577859 Chr.19: 14743254-14743254 on Build GRCh38
rs1001165342 Chr.19: 14743365-14743365 on Build GRCh38
rs914038712 Chr.19: 14743297-14743297 on Build GRCh38
rs202101555 Chr.19: 14743420-14743420 on Build GRCh38
rs749460881 Chr.19: 14743414-14743414 on Build GRCh38

View Full Product Details