Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 6088918-6089143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FERMT1
Entrez Gene ID
55612
Gene Name
fermitin family member 1
Gene Aliases
C20orf42, DTGCU2, KIND1, UNC112A, URP1
UniGene
Hs.472054
Gene Chromosome Location
Chr.20: 6074845-6123544 on Build GRCh38
Transcripts
NM_017671.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
478364 c.1181T>G p.F394C Chr.20: 6089048-6089048 on Build GRCh38
1713154 c.1199C>T p.S400F Chr.20: 6089030-6089030 on Build GRCh38
3548577 c.1185C>T p.I395I Chr.20: 6089044-6089044 on Build GRCh38
3673152 c.1222G>T p.E408* Chr.20: 6089007-6089007 on Build GRCh38
4684341 c.1205C>T p.A402V Chr.20: 6089024-6089024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927239737 Chr.20: 6089065-6089065 on Build GRCh38
rs746370433 Chr.20: 6088988-6088988 on Build GRCh38
rs770125798 Chr.20: 6088990-6088990 on Build GRCh38
rs869312718 Chr.20: 6089031-6089031 on Build GRCh38
rs374069028 Chr.20: 6089062-6089062 on Build GRCh38
rs778100302 Chr.20: 6089080-6089080 on Build GRCh38
rs373454595 Chr.20: 6089111-6089111 on Build GRCh38
rs376957545 Chr.20: 6089115-6089115 on Build GRCh38
rs869312729 Chr.20: 6089068-6089068 on Build GRCh38
rs766950162 Chr.20: 6089046-6089046 on Build GRCh38
rs749615948 Chr.20: 6089105-6089105 on Build GRCh38
rs780581206 Chr.20: 6089101-6089101 on Build GRCh38
rs536285387 Chr.20: 6089081-6089081 on Build GRCh38
rs773939268 Chr.20: 6089113-6089113 on Build GRCh38
rs757299220 Chr.20: 6089085-6089085 on Build GRCh38
rs869312730 Chr.20: 6089053-6089053 on Build GRCh38
rs781688885 Chr.20: 6088967-6088967 on Build GRCh38
rs746023395 Chr.20: 6089088-6089088 on Build GRCh38
rs753662479 Chr.20: 6089068-6089068 on Build GRCh38
rs770269738 Chr.20: 6089094-6089094 on Build GRCh38
rs761341202 Chr.20: 6089020-6089020 on Build GRCh38
rs139181861 Chr.20: 6089002-6089002 on Build GRCh38
rs369591144 Chr.20: 6088998-6088998 on Build GRCh38
rs781132211 Chr.20: 6089087-6089087 on Build GRCh38
rs768269556 Chr.20: 6089004-6089004 on Build GRCh38
rs750308235 Chr.20: 6089051-6089051 on Build GRCh38
rs111975159 Chr.20: 6088951-6088951 on Build GRCh38
rs35413391 Chr.20: 6089076-6089076 on Build GRCh38

View Full Product Details