Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3867869-3868142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DFFB
Entrez Gene ID
1677
Gene Name
DNA fragmentation factor subunit beta
Gene Aliases
CAD, CPAN, DFF-40, DFF2, DFF40
UniGene
Hs.133089
Gene Chromosome Location
Chr.1: 3857143-3885429 on Build GRCh38
Transcripts
NM_001282669.1 NM_001320132.1 NM_001320136.1 NM_004402.3 NR_104222.1 NR_135150.1 NR_135151.1 NR_135152.1 XM_017000498.1 XM_017000499.1 XM_017000500.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7262659 c.487C>T p.R163W Chr.1: 3868030-3868030 on Build GRCh38
350048 c.489G>T p.R163R Chr.1: 3868032-3868032 on Build GRCh38
5820902 c.431-2A>T p.? Chr.1: 3867972-3867972 on Build GRCh38
1203415 c.443G>A p.R148Q Chr.1: 3867986-3867986 on Build GRCh38
6642771 c.493C>T p.R165W Chr.1: 3868036-3868036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370493706 Chr.1: 3868085-3868085 on Build GRCh38
rs762598482 Chr.1: 3868041-3868041 on Build GRCh38
rs761503584 Chr.1: 3868049-3868049 on Build GRCh38
rs750391864 Chr.1: 3867986-3867986 on Build GRCh38
rs751793100 Chr.1: 3868098-3868098 on Build GRCh38
rs910936059 Chr.1: 3867950-3867950 on Build GRCh38
rs200175328 Chr.1: 3867951-3867951 on Build GRCh38
rs371788433 Chr.1: 3867985-3867985 on Build GRCh38
rs751231800 Chr.1: 3867968-3867968 on Build GRCh38
rs776032198 Chr.1: 3868040-3868040 on Build GRCh38
rs768264214 Chr.1: 3868044-3868044 on Build GRCh38
rs763876210 Chr.1: 3867961-3867961 on Build GRCh38
rs200707180 Chr.1: 3868086-3868086 on Build GRCh38
rs755958810 Chr.1: 3867997-3867997 on Build GRCh38
rs760490486 Chr.1: 3868073-3868073 on Build GRCh38
rs377219792 Chr.1: 3868084-3868084 on Build GRCh38
rs1010089955 Chr.1: 3867910-3867910 on Build GRCh38
rs750147209 Chr.1: 3868072-3868072 on Build GRCh38
rs758451503 Chr.1: 3868023-3868023 on Build GRCh38
rs771289652 Chr.1: 3868031-3868031 on Build GRCh38
rs916342696 Chr.1: 3867977-3867977 on Build GRCh38
rs199677789 Chr.1: 3867959-3867959 on Build GRCh38
rs72848469 Chr.1: 3867900-3867900 on Build GRCh38
rs373643674 Chr.1: 3868017-3868017 on Build GRCh38
rs766278184 Chr.1: 3868074-3868074 on Build GRCh38
rs758862022 Chr.1: 3868048-3868048 on Build GRCh38
rs370082825 Chr.1: 3867953-3867953 on Build GRCh38
rs771273194 Chr.1: 3867934-3867934 on Build GRCh38
rs201973418 Chr.1: 3868037-3868037 on Build GRCh38
rs757421931 Chr.1: 3868104-3868104 on Build GRCh38
rs752761449 Chr.1: 3868005-3868005 on Build GRCh38
rs531823030 Chr.1: 3867918-3867918 on Build GRCh38
rs76754622 Chr.1: 3867890-3867890 on Build GRCh38
rs761754804 Chr.1: 3867969-3867969 on Build GRCh38
rs767370905 Chr.1: 3868056-3868056 on Build GRCh38
rs903732277 Chr.1: 3868042-3868042 on Build GRCh38
rs376189053 Chr.1: 3867939-3867939 on Build GRCh38
rs114770404 Chr.1: 3867960-3867960 on Build GRCh38
rs987047223 Chr.1: 3867928-3867928 on Build GRCh38
rs747424032 Chr.1: 3867927-3867927 on Build GRCh38
rs779084705 Chr.1: 3867999-3867999 on Build GRCh38
rs751383491 Chr.1: 3867949-3867949 on Build GRCh38
rs778028989 Chr.1: 3868027-3868027 on Build GRCh38
rs777261791 Chr.1: 3867947-3867947 on Build GRCh38
rs984965593 Chr.1: 3867905-3867905 on Build GRCh38
rs1020660505 Chr.1: 3867923-3867923 on Build GRCh38
rs747227784 Chr.1: 3868030-3868030 on Build GRCh38
rs746407986 Chr.1: 3868036-3868036 on Build GRCh38
rs762092035 Chr.1: 3867907-3867907 on Build GRCh38
rs781353590 Chr.1: 3868032-3868032 on Build GRCh38
rs376710097 Chr.1: 3868114-3868114 on Build GRCh38
rs534787656 Chr.1: 3868089-3868089 on Build GRCh38

View Full Product Details