Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25209748-25210019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD6
Entrez Gene ID
955
Gene Name
ectonucleoside triphosphate diphosphohydrolase 6 (putative)
Gene Aliases
CD39L2, IL-6SAG, IL6ST2, NTPDase-6, dJ738P15.3
UniGene
Hs.500375
Gene Chromosome Location
Chr.20: 25195670-25228075 on Build GRCh38
Transcripts
NM_001114089.3 NM_001247.4 NM_001317941.2 NM_001322378.1 NM_001322379.1 NM_001322380.1 NM_001322381.1 NM_001322382.1 NM_001322383.1 NM_001322384.1 NM_001322385.1 NM_001322386.1 NM_001322387.1 NM_001322388.1 NM_001322389.1 NM_001322390.1 NM_001322391.1 NM_001322392.1 NM_001322393.1 NM_001322394.1 NM_001322395.1 NM_001322396.1 NM_001322397.1 NM_001322398.1 XM_006723665.2 XM_011529396.1 XM_011529397.1 XM_017028129.1 XM_017028130.1 XM_017028131.1 XM_017028132.1 XM_017028133.1 XM_017028134.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4491618 c.383C>T p.P128L Chr.20: 25209855-25209855 on Build GRCh38
5041178 c.395A>G p.H132R Chr.20: 25209867-25209867 on Build GRCh38
1025319 c.437C>T p.A146V Chr.20: 25209909-25209909 on Build GRCh38
6654336 c.396C>T p.H132H Chr.20: 25209868-25209868 on Build GRCh38
6525710 c.414G>A p.L138L Chr.20: 25209886-25209886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772057782 Chr.20: 25209856-25209856 on Build GRCh38
rs201877644 Chr.20: 25209959-25209959 on Build GRCh38
rs144718324 Chr.20: 25209869-25209869 on Build GRCh38
rs746887770 Chr.20: 25209859-25209859 on Build GRCh38
rs779053618 Chr.20: 25209846-25209846 on Build GRCh38
rs770945981 Chr.20: 25209977-25209977 on Build GRCh38
rs112981620 Chr.20: 25209874-25209874 on Build GRCh38
rs980220876 Chr.20: 25209780-25209780 on Build GRCh38
rs749202051 Chr.20: 25209925-25209930 on Build GRCh38
rs576723642 Chr.20: 25209784-25209784 on Build GRCh38
rs755050693 Chr.20: 25209942-25209942 on Build GRCh38
rs1048919070 Chr.20: 25209998-25209998 on Build GRCh38
rs371406615 Chr.20: 25209818-25209818 on Build GRCh38
rs750983984 Chr.20: 25209801-25209801 on Build GRCh38
rs762045177 Chr.20: 25209904-25209904 on Build GRCh38
rs903187943 Chr.20: 25209987-25209987 on Build GRCh38
rs767809268 Chr.20: 25209916-25209916 on Build GRCh38
rs749355416 Chr.20: 25209834-25209834 on Build GRCh38
rs747942843 Chr.20: 25209855-25209855 on Build GRCh38
rs575862145 Chr.20: 25209907-25209907 on Build GRCh38
rs768733518 Chr.20: 25209937-25209949 on Build GRCh38
rs113444643 Chr.20: 25209887-25209887 on Build GRCh38
rs1007707 Chr.20: 25209962-25209962 on Build GRCh38
rs756570568 Chr.20: 25209931-25209931 on Build GRCh38
rs200908979 Chr.20: 25209844-25209844 on Build GRCh38
rs573910558 Chr.20: 25209868-25209868 on Build GRCh38
rs758329576 Chr.20: 25209804-25209804 on Build GRCh38
rs1044567 Chr.20: 25209884-25209884 on Build GRCh38
rs545735578 Chr.20: 25209858-25209858 on Build GRCh38
rs375548383 Chr.20: 25209969-25209969 on Build GRCh38
rs758710121 Chr.20: 25209961-25209961 on Build GRCh38
rs779190150 Chr.20: 25209943-25209943 on Build GRCh38
rs768200669 Chr.20: 25209967-25209968 on Build GRCh38
rs758994846 Chr.20: 25209883-25209883 on Build GRCh38
rs746832636 Chr.20: 25209972-25209972 on Build GRCh38
rs772025776 Chr.20: 25209922-25209922 on Build GRCh38
rs773707276 Chr.20: 25209939-25209939 on Build GRCh38
rs541645883 Chr.20: 25209941-25209941 on Build GRCh38
rs1036483520 Chr.20: 25209776-25209776 on Build GRCh38
rs775048872 Chr.20: 25209894-25209894 on Build GRCh38
rs756624197 Chr.20: 25209812-25209812 on Build GRCh38
rs367662355 Chr.20: 25209811-25209811 on Build GRCh38
rs766817176 Chr.20: 25209935-25209935 on Build GRCh38
rs750786416 Chr.20: 25209928-25209928 on Build GRCh38
rs374001806 Chr.20: 25209957-25209957 on Build GRCh38

View Full Product Details