Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206647938-206648211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK3
Entrez Gene ID
8444
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 3
Gene Aliases
DYRK5, RED, REDK, hYAK3-2
UniGene
Hs.164267
Gene Chromosome Location
Chr.1: 206635536-206655158 on Build GRCh38
Transcripts
NM_001004023.1 NM_003582.2 XM_005273315.4 XM_011510061.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1583995 c.908T>C p.L303S Chr.1: 206648166-206648166 on Build GRCh38
5911304 c.765G>T p.L255F Chr.1: 206648068-206648068 on Build GRCh38
1668421 c.871C>A p.L291I Chr.1: 206648174-206648174 on Build GRCh38
3482464 c.769A>T p.S257C Chr.1: 206648027-206648027 on Build GRCh38
3482463 c.829A>T p.S277C Chr.1: 206648027-206648027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782411067 Chr.1: 206648038-206648038 on Build GRCh38
rs782053343 Chr.1: 206648085-206648085 on Build GRCh38
rs923464924 Chr.1: 206648077-206648077 on Build GRCh38
rs782636809 Chr.1: 206648147-206648147 on Build GRCh38
rs367696416 Chr.1: 206648121-206648121 on Build GRCh38
rs199694885 Chr.1: 206648131-206648131 on Build GRCh38
rs782269982 Chr.1: 206648035-206648035 on Build GRCh38
rs201468157 Chr.1: 206648025-206648025 on Build GRCh38
rs371479482 Chr.1: 206648125-206648125 on Build GRCh38
rs782388988 Chr.1: 206648064-206648064 on Build GRCh38
rs556178304 Chr.1: 206648122-206648122 on Build GRCh38
rs199718118 Chr.1: 206647988-206647988 on Build GRCh38
rs560144939 Chr.1: 206648050-206648050 on Build GRCh38
rs782438854 Chr.1: 206648013-206648013 on Build GRCh38
rs782390001 Chr.1: 206647961-206647961 on Build GRCh38
rs202193987 Chr.1: 206647962-206647962 on Build GRCh38
rs1035558323 Chr.1: 206648146-206648146 on Build GRCh38
rs782819674 Chr.1: 206648016-206648016 on Build GRCh38
rs781940613 Chr.1: 206648155-206648155 on Build GRCh38
rs55766763 Chr.1: 206648018-206648018 on Build GRCh38
rs782572211 Chr.1: 206648039-206648039 on Build GRCh38
rs143076249 Chr.1: 206648161-206648161 on Build GRCh38
rs782184680 Chr.1: 206648040-206648040 on Build GRCh38
rs201551415 Chr.1: 206648130-206648130 on Build GRCh38
rs200128484 Chr.1: 206648175-206648175 on Build GRCh38
rs781936999 Chr.1: 206647974-206647974 on Build GRCh38
rs782369987 Chr.1: 206648165-206648165 on Build GRCh38
rs782510096 Chr.1: 206648026-206648026 on Build GRCh38
rs200513273 Chr.1: 206648135-206648135 on Build GRCh38
rs201402787 Chr.1: 206648046-206648046 on Build GRCh38
rs782345381 Chr.1: 206648047-206648047 on Build GRCh38
rs782274145 Chr.1: 206647975-206647975 on Build GRCh38
rs782675715 Chr.1: 206647960-206647961 on Build GRCh38
rs782034506 Chr.1: 206648075-206648075 on Build GRCh38
rs781823889 Chr.1: 206648091-206648091 on Build GRCh38
rs186754669 Chr.1: 206648008-206648008 on Build GRCh38
rs781850677 Chr.1: 206648009-206648009 on Build GRCh38
rs199522696 Chr.1: 206648096-206648096 on Build GRCh38
rs782305912 Chr.1: 206647972-206647972 on Build GRCh38
rs782082612 Chr.1: 206647997-206647997 on Build GRCh38
rs782146559 Chr.1: 206648011-206648011 on Build GRCh38
rs782299855 Chr.1: 206648137-206648137 on Build GRCh38
rs781791751 Chr.1: 206648017-206648017 on Build GRCh38
rs782387740 Chr.1: 206648107-206648108 on Build GRCh38
rs55982485 Chr.1: 206648180-206648180 on Build GRCh38
rs200692265 Chr.1: 206648103-206648103 on Build GRCh38
rs781783476 Chr.1: 206648123-206648123 on Build GRCh38
rs373891264 Chr.1: 206648052-206648052 on Build GRCh38
rs55726720 Chr.1: 206648089-206648089 on Build GRCh38
rs782645630 Chr.1: 206648033-206648033 on Build GRCh38
rs182757711 Chr.1: 206647971-206647971 on Build GRCh38

View Full Product Details