Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 81992635-81992878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA31D1
Entrez Gene ID
389763
Gene Name
SPATA31 subfamily D member 1
Gene Aliases
FAM75D1
UniGene
Hs.209545
Gene Chromosome Location
Chr.9: 81987404-81995256 on Build GRCh38
Transcripts
NM_001001670.2 XM_017014710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1110479 c.2366C>T p.P789L Chr.9: 81992836-81992836 on Build GRCh38
6013517 c.2309G>A p.G770E Chr.9: 81992779-81992779 on Build GRCh38
4139609 c.2363G>T p.G788V Chr.9: 81992833-81992833 on Build GRCh38
3716305 c.2342C>G p.P781R Chr.9: 81992812-81992812 on Build GRCh38
5664313 c.2259T>G p.P753P Chr.9: 81992729-81992729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187675943 Chr.9: 81992663-81992663 on Build GRCh38
rs753234214 Chr.9: 81992734-81992734 on Build GRCh38
rs371028145 Chr.9: 81992818-81992818 on Build GRCh38
rs191755245 Chr.9: 81992714-81992714 on Build GRCh38
rs751252513 Chr.9: 81992782-81992782 on Build GRCh38
rs777773457 Chr.9: 81992692-81992692 on Build GRCh38
rs769681115 Chr.9: 81992827-81992827 on Build GRCh38
rs907560592 Chr.9: 81992674-81992674 on Build GRCh38
rs767217256 Chr.9: 81992795-81992795 on Build GRCh38
rs774576065 Chr.9: 81992698-81992698 on Build GRCh38
rs758482828 Chr.9: 81992819-81992819 on Build GRCh38
rs781480536 Chr.9: 81992728-81992728 on Build GRCh38
rs188127937 Chr.9: 81992810-81992810 on Build GRCh38
rs565458037 Chr.9: 81992821-81992821 on Build GRCh38
rs776231374 Chr.9: 81992772-81992772 on Build GRCh38
rs201213048 Chr.9: 81992684-81992684 on Build GRCh38
rs575202702 Chr.9: 81992662-81992662 on Build GRCh38
rs376249496 Chr.9: 81992833-81992833 on Build GRCh38
rs768480958 Chr.9: 81992769-81992769 on Build GRCh38
rs138409860 Chr.9: 81992778-81992778 on Build GRCh38
rs200441343 Chr.9: 81992673-81992673 on Build GRCh38
rs373040016 Chr.9: 81992830-81992830 on Build GRCh38
rs773945588 Chr.9: 81992836-81992836 on Build GRCh38
rs369132919 Chr.9: 81992741-81992741 on Build GRCh38
rs754380091 Chr.9: 81992809-81992809 on Build GRCh38
rs369084497 Chr.9: 81992696-81992696 on Build GRCh38
rs759323799 Chr.9: 81992794-81992794 on Build GRCh38
rs369693916 Chr.9: 81992853-81992853 on Build GRCh38
rs374159797 Chr.9: 81992678-81992678 on Build GRCh38
rs750073015 Chr.9: 81992661-81992661 on Build GRCh38
rs748647179 Chr.9: 81992681-81992681 on Build GRCh38
rs545600287 Chr.9: 81992727-81992727 on Build GRCh38
rs377116387 Chr.9: 81992672-81992672 on Build GRCh38
rs1027176653 Chr.9: 81992852-81992852 on Build GRCh38
rs755419107 Chr.9: 81992670-81992670 on Build GRCh38
rs764583739 Chr.9: 81992845-81992845 on Build GRCh38
rs34402732 Chr.9: 81992704-81992705 on Build GRCh38
rs779018477 Chr.9: 81992811-81992811 on Build GRCh38
rs775232690 Chr.9: 81992841-81992841 on Build GRCh38
rs768093242 Chr.9: 81992787-81992787 on Build GRCh38
rs368073850 Chr.9: 81992764-81992764 on Build GRCh38
rs749265620 Chr.9: 81992726-81992726 on Build GRCh38
rs183138674 Chr.9: 81992780-81992780 on Build GRCh38
rs756521351 Chr.9: 81992737-81992737 on Build GRCh38
rs374062634 Chr.9: 81992846-81992846 on Build GRCh38
rs372215050 Chr.9: 81992837-81992837 on Build GRCh38
rs762580342 Chr.9: 81992721-81992721 on Build GRCh38
rs560197350 Chr.9: 81992699-81992699 on Build GRCh38
rs764484112 Chr.9: 81992805-81992805 on Build GRCh38
rs779075552 Chr.9: 81992750-81992750 on Build GRCh38
rs34942699 Chr.9: 81992776-81992777 on Build GRCh38
rs768478334 Chr.9: 81992700-81992700 on Build GRCh38
rs761672079 Chr.9: 81992775-81992775 on Build GRCh38
rs756535158 Chr.9: 81992804-81992804 on Build GRCh38
rs778349299 Chr.9: 81992739-81992739 on Build GRCh38
rs751935060 Chr.9: 81992668-81992668 on Build GRCh38
rs752941119 Chr.9: 81992796-81992796 on Build GRCh38
rs200798404 Chr.9: 81992715-81992715 on Build GRCh38
rs749832884 Chr.9: 81992719-81992719 on Build GRCh38
rs10867826 Chr.9: 81992843-81992843 on Build GRCh38

View Full Product Details