Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 183940428-183940653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLGALT2
Entrez Gene ID
23127
Gene Name
collagen beta(1-O)galactosyltransferase 2
Gene Aliases
C1orf17, GLT25D2
UniGene
Hs.387995
Gene Chromosome Location
Chr.1: 183929662-184051612 on Build GRCh38
Transcripts
NM_001303420.1 NM_001303421.1 NM_015101.3 XM_011509332.2 XM_011509334.2 XM_011509335.2 XM_011509336.2 XM_011509337.2 XM_011509338.2 XM_017000750.1 XM_017000751.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4468440 c.1544C>T p.P515L Chr.1: 183940641-183940641 on Build GRCh38
4025960 c.792A>G p.P264P Chr.1: 183940604-183940604 on Build GRCh38
6508369 c.1554G>T p.K518N Chr.1: 183940631-183940631 on Build GRCh38
4468441 c.755C>T p.P252L Chr.1: 183940641-183940641 on Build GRCh38
3750692 c.1602C>T p.P534P Chr.1: 183940583-183940583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043275657 Chr.1: 183940457-183940457 on Build GRCh38
rs754162801 Chr.1: 183940628-183940628 on Build GRCh38
rs772059612 Chr.1: 183940546-183940546 on Build GRCh38
rs767583258 Chr.1: 183940602-183940602 on Build GRCh38
rs1054749589 Chr.1: 183940606-183940606 on Build GRCh38
rs189112153 Chr.1: 183940488-183940488 on Build GRCh38
rs904281373 Chr.1: 183940468-183940468 on Build GRCh38
rs764486932 Chr.1: 183940599-183940599 on Build GRCh38
rs2296713 Chr.1: 183940583-183940583 on Build GRCh38
rs2296712 Chr.1: 183940499-183940499 on Build GRCh38
rs750483975 Chr.1: 183940616-183940616 on Build GRCh38
rs753279163 Chr.1: 183940590-183940592 on Build GRCh38
rs895512568 Chr.1: 183940497-183940497 on Build GRCh38
rs952790999 Chr.1: 183940557-183940557 on Build GRCh38
rs773261512 Chr.1: 183940551-183940551 on Build GRCh38
rs778710803 Chr.1: 183940534-183940534 on Build GRCh38
rs368052719 Chr.1: 183940545-183940545 on Build GRCh38
rs770436368 Chr.1: 183940579-183940579 on Build GRCh38
rs932540996 Chr.1: 183940454-183940454 on Build GRCh38
rs747960853 Chr.1: 183940538-183940538 on Build GRCh38
rs934374811 Chr.1: 183940483-183940483 on Build GRCh38
rs769313060 Chr.1: 183940627-183940627 on Build GRCh38
rs767448919 Chr.1: 183940539-183940540 on Build GRCh38
rs905433970 Chr.1: 183940527-183940527 on Build GRCh38
rs139035282 Chr.1: 183940489-183940489 on Build GRCh38
rs185200430 Chr.1: 183940558-183940558 on Build GRCh38
rs776028862 Chr.1: 183940580-183940580 on Build GRCh38
rs193133730 Chr.1: 183940513-183940513 on Build GRCh38
rs757732508 Chr.1: 183940601-183940601 on Build GRCh38
rs758236512 Chr.1: 183940631-183940631 on Build GRCh38
rs775193430 Chr.1: 183940588-183940588 on Build GRCh38
rs200271185 Chr.1: 183940533-183940533 on Build GRCh38
rs761020830 Chr.1: 183940618-183940618 on Build GRCh38
rs964293528 Chr.1: 183940512-183940512 on Build GRCh38
rs140712886 Chr.1: 183940594-183940594 on Build GRCh38
rs542850872 Chr.1: 183940466-183940466 on Build GRCh38
rs1025262617 Chr.1: 183940517-183940517 on Build GRCh38
rs759172474 Chr.1: 183940582-183940582 on Build GRCh38
rs756293963 Chr.1: 183940619-183940619 on Build GRCh38
rs374917733 Chr.1: 183940600-183940600 on Build GRCh38
rs746060055 Chr.1: 183940530-183940530 on Build GRCh38
rs368161881 Chr.1: 183940623-183940623 on Build GRCh38

View Full Product Details