Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 15215925-15216184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM171A1
Entrez Gene ID
221061
Gene Name
family with sequence similarity 171 member A1
Gene Aliases
C10orf38
UniGene
Hs.66762
Gene Chromosome Location
Chr.10: 15211643-15374532 on Build GRCh38
Transcripts
NM_001010924.1 XM_011519378.2 XM_017015904.1 XM_017015905.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4013032 c.922G>A p.A308T Chr.10: 15216060-15216060 on Build GRCh38
3699871 c.877G>A p.V293I Chr.10: 15216105-15216105 on Build GRCh38
1728924 c.907A>T p.T303S Chr.10: 15216075-15216075 on Build GRCh38
916703 c.961C>A p.L321M Chr.10: 15216021-15216021 on Build GRCh38
6658877 c.972T>C p.L324L Chr.10: 15216010-15216010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763322122 Chr.10: 15215975-15215975 on Build GRCh38
rs767615993 Chr.10: 15216028-15216028 on Build GRCh38
rs572984970 Chr.10: 15216130-15216130 on Build GRCh38
rs751813281 Chr.10: 15215988-15215988 on Build GRCh38
rs977585806 Chr.10: 15215970-15215970 on Build GRCh38
rs923721593 Chr.10: 15216013-15216013 on Build GRCh38
rs754847829 Chr.10: 15216074-15216074 on Build GRCh38
rs765730571 Chr.10: 15215968-15215968 on Build GRCh38
rs764253594 Chr.10: 15215984-15215984 on Build GRCh38
rs142266275 Chr.10: 15216118-15216118 on Build GRCh38
rs1042535557 Chr.10: 15216141-15216141 on Build GRCh38
rs777331323 Chr.10: 15216106-15216106 on Build GRCh38
rs764907660 Chr.10: 15216132-15216132 on Build GRCh38
rs747778479 Chr.10: 15216082-15216082 on Build GRCh38
rs546058977 Chr.10: 15216094-15216094 on Build GRCh38
rs867917252 Chr.10: 15216066-15216066 on Build GRCh38
rs377642428 Chr.10: 15216058-15216058 on Build GRCh38
rs779864841 Chr.10: 15216063-15216063 on Build GRCh38
rs775858297 Chr.10: 15215973-15215973 on Build GRCh38
rs144702270 Chr.10: 15216002-15216002 on Build GRCh38
rs140163568 Chr.10: 15216073-15216073 on Build GRCh38
rs1001151223 Chr.10: 15216158-15216158 on Build GRCh38
rs750530629 Chr.10: 15216057-15216057 on Build GRCh38
rs778889217 Chr.10: 15216076-15216076 on Build GRCh38
rs369414724 Chr.10: 15216155-15216155 on Build GRCh38
rs949593861 Chr.10: 15216087-15216087 on Build GRCh38

View Full Product Details