Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 2175709-2175982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMDS
Entrez Gene ID
2762
Gene Name
GDP-mannose 4,6-dehydratase
Gene Aliases
GMD, SDR3E1
UniGene
Hs.144496
Gene Chromosome Location
Chr.6: 1623800-2245634 on Build GRCh38
Transcripts
NM_001253846.1 NM_001500.3 XM_006715066.2 XM_011514500.1 XM_011514502.2 XM_011514503.2 XM_011514505.1 XM_011514506.1 XM_011514507.1

SNP Detail

SNP ID Chromosome Location
rs992651428 Chr.6: 2175877-2175877 on Build GRCh38
rs895017361 Chr.6: 2175867-2175867 on Build GRCh38
rs546194754 Chr.6: 2175844-2175844 on Build GRCh38
rs541303539 Chr.6: 2175810-2175810 on Build GRCh38
rs907960568 Chr.6: 2175845-2175845 on Build GRCh38
rs1031850973 Chr.6: 2175750-2175750 on Build GRCh38
rs974992633 Chr.6: 2175821-2175821 on Build GRCh38
rs1054882702 Chr.6: 2175856-2175856 on Build GRCh38
rs757836677 Chr.6: 2175925-2175925 on Build GRCh38
rs373096583 Chr.6: 2175842-2175842 on Build GRCh38
rs764767887 Chr.6: 2175746-2175746 on Build GRCh38
rs1017809080 Chr.6: 2175740-2175740 on Build GRCh38
rs1008982307 Chr.6: 2175776-2175776 on Build GRCh38
rs145498101 Chr.6: 2175800-2175800 on Build GRCh38
rs952329204 Chr.6: 2175826-2175826 on Build GRCh38
rs752322217 Chr.6: 2175749-2175749 on Build GRCh38
rs543248059 Chr.6: 2175814-2175814 on Build GRCh38
rs549176638 Chr.6: 2175808-2175808 on Build GRCh38
rs199885372 Chr.6: 2175835-2175835 on Build GRCh38
rs760965010 Chr.6: 2175960-2175960 on Build GRCh38
rs1043611700 Chr.6: 2175943-2175943 on Build GRCh38
rs139589788 Chr.6: 2175955-2175955 on Build GRCh38
rs939414649 Chr.6: 2175875-2175875 on Build GRCh38
rs189686901 Chr.6: 2175822-2175822 on Build GRCh38
rs939462840 Chr.6: 2175831-2175831 on Build GRCh38
rs149671078 Chr.6: 2175764-2175764 on Build GRCh38

View Full Product Details