Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 71026564-71026825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCK
Entrez Gene ID
1633
Gene Name
deoxycytidine kinase
Gene Aliases
-
UniGene
Hs.709
Gene Chromosome Location
Chr.4: 70993548-71030914 on Build GRCh38
Transcripts
NM_000788.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3993769 c.685C>A p.Q229K Chr.4: 71026684-71026684 on Build GRCh38
195629 c.675C>T p.F225F Chr.4: 71026674-71026674 on Build GRCh38
268984 c.667A>G p.T223A Chr.4: 71026666-71026666 on Build GRCh38
6447064 c.692T>C p.V231A Chr.4: 71026691-71026691 on Build GRCh38
4968107 c.676G>A p.D226N Chr.4: 71026675-71026675 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745977177 Chr.4: 71026736-71026736 on Build GRCh38
rs781389066 Chr.4: 71026731-71026731 on Build GRCh38
rs374531654 Chr.4: 71026777-71026777 on Build GRCh38
rs1043095786 Chr.4: 71026596-71026596 on Build GRCh38
rs780722524 Chr.4: 71026745-71026745 on Build GRCh38
rs781335222 Chr.4: 71026764-71026767 on Build GRCh38
rs761353817 Chr.4: 71026785-71026785 on Build GRCh38
rs778833527 Chr.4: 71026716-71026716 on Build GRCh38
rs890347494 Chr.4: 71026630-71026630 on Build GRCh38
rs749599213 Chr.4: 71026747-71026747 on Build GRCh38
rs200315482 Chr.4: 71026675-71026675 on Build GRCh38
rs1486271 Chr.4: 71026796-71026796 on Build GRCh38
rs768765506 Chr.4: 71026615-71026615 on Build GRCh38
rs773882928 Chr.4: 71026658-71026658 on Build GRCh38
rs369967087 Chr.4: 71026726-71026726 on Build GRCh38
rs972568532 Chr.4: 71026789-71026789 on Build GRCh38
rs766322484 Chr.4: 71026696-71026696 on Build GRCh38
rs1008569242 Chr.4: 71026634-71026634 on Build GRCh38
rs564522050 Chr.4: 71026674-71026674 on Build GRCh38
rs770375633 Chr.4: 71026744-71026744 on Build GRCh38
rs763434371 Chr.4: 71026623-71026623 on Build GRCh38
rs747104239 Chr.4: 71026614-71026615 on Build GRCh38
rs996095862 Chr.4: 71026629-71026629 on Build GRCh38
rs754730723 Chr.4: 71026708-71026708 on Build GRCh38
rs368766396 Chr.4: 71026680-71026680 on Build GRCh38
rs904625188 Chr.4: 71026625-71026625 on Build GRCh38
rs1031607418 Chr.4: 71026617-71026617 on Build GRCh38
rs961170674 Chr.4: 71026786-71026786 on Build GRCh38
rs751716190 Chr.4: 71026725-71026725 on Build GRCh38
rs771801486 Chr.4: 71026787-71026787 on Build GRCh38
rs768900840 Chr.4: 71026758-71026758 on Build GRCh38
rs760561814 Chr.4: 71026693-71026693 on Build GRCh38
rs879053912 Chr.4: 71026622-71026622 on Build GRCh38
rs1019426207 Chr.4: 71026784-71026784 on Build GRCh38
rs775957733 Chr.4: 71026620-71026620 on Build GRCh38
rs768299319 Chr.4: 71026635-71026635 on Build GRCh38

View Full Product Details