Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 160199024-160199289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FABP6
Entrez Gene ID
2172
Gene Name
fatty acid binding protein 6
Gene Aliases
I-15P, I-BABP, I-BALB, I-BAP, ILBP, ILBP3, ILLBP
UniGene
Hs.519719
Gene Chromosome Location
Chr.5: 160187367-160238722 on Build GRCh38
Transcripts
NM_001040442.1 NM_001130958.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5141494 c.22A>G p.M8V Chr.5: 160199128-160199128 on Build GRCh38
6436344 c.44T>A p.L15Q Chr.5: 160199150-160199150 on Build GRCh38
3613420 c.49C>A p.Q17K Chr.5: 160199155-160199155 on Build GRCh38
7148431 c.31G>A p.E11K Chr.5: 160199137-160199137 on Build GRCh38
6657894 c.41C>T p.A14V Chr.5: 160199147-160199147 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750540506 Chr.5: 160199085-160199085 on Build GRCh38
rs777059808 Chr.5: 160199088-160199095 on Build GRCh38
rs774753160 Chr.5: 160199173-160199173 on Build GRCh38
rs765185372 Chr.5: 160199079-160199079 on Build GRCh38
rs767724967 Chr.5: 160199187-160199187 on Build GRCh38
rs768035620 Chr.5: 160199095-160199095 on Build GRCh38
rs751957245 Chr.5: 160199091-160199091 on Build GRCh38
rs763197384 Chr.5: 160199088-160199088 on Build GRCh38
rs759386302 Chr.5: 160199062-160199062 on Build GRCh38
rs755346751 Chr.5: 160199092-160199092 on Build GRCh38
rs756576198 Chr.5: 160199201-160199201 on Build GRCh38
rs775327708 Chr.5: 160199249-160199249 on Build GRCh38
rs766772386 Chr.5: 160199090-160199090 on Build GRCh38
rs550873727 Chr.5: 160199175-160199175 on Build GRCh38
rs774130643 Chr.5: 160199060-160199060 on Build GRCh38
rs138753107 Chr.5: 160199240-160199240 on Build GRCh38
rs1011174407 Chr.5: 160199252-160199252 on Build GRCh38
rs771727828 Chr.5: 160199122-160199122 on Build GRCh38
rs753111139 Chr.5: 160199199-160199199 on Build GRCh38
rs1021180918 Chr.5: 160199260-160199260 on Build GRCh38
rs753104712 Chr.5: 160199103-160199103 on Build GRCh38
rs146032989 Chr.5: 160199121-160199121 on Build GRCh38
rs66840636 Chr.5: 160199237-160199237 on Build GRCh38
rs1054124789 Chr.5: 160199247-160199247 on Build GRCh38
rs202217927 Chr.5: 160199132-160199132 on Build GRCh38
rs760561971 Chr.5: 160199261-160199261 on Build GRCh38
rs760701869 Chr.5: 160199073-160199073 on Build GRCh38
rs756666320 Chr.5: 160199105-160199105 on Build GRCh38
rs199868205 Chr.5: 160199131-160199131 on Build GRCh38
rs775331841 Chr.5: 160199068-160199068 on Build GRCh38
rs748865194 Chr.5: 160199112-160199112 on Build GRCh38
rs531102670 Chr.5: 160199167-160199167 on Build GRCh38
rs768640444 Chr.5: 160199146-160199146 on Build GRCh38
rs770574469 Chr.5: 160199057-160199057 on Build GRCh38
rs773418482 Chr.5: 160199148-160199148 on Build GRCh38
rs746860569 Chr.5: 160199134-160199134 on Build GRCh38
rs774979117 Chr.5: 160199100-160199100 on Build GRCh38
rs772026716 Chr.5: 160199066-160199066 on Build GRCh38
rs201223363 Chr.5: 160199120-160199120 on Build GRCh38
rs777082611 Chr.5: 160199106-160199106 on Build GRCh38
rs760036322 Chr.5: 160199117-160199118 on Build GRCh38

View Full Product Details