Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 93678886-93679111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRADD
Entrez Gene ID
8738
Gene Name
CASP2 and RIPK1 domain containing adaptor with death domain
Gene Aliases
MRT34, RAIDD
UniGene
Hs.38533
Gene Chromosome Location
Chr.12: 93677342-93894840 on Build GRCh38
Transcripts
NM_001320099.1 NM_001320100.1 NM_001320101.1 NM_003805.4 NR_135147.1 XM_017020142.1 XM_017020143.1 XM_017020144.1 XM_017020145.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
347015 c.136G>T p.A46S Chr.12: 93678910-93678910 on Build GRCh38
5478408 c.286G>A p.D96N Chr.12: 93679060-93679060 on Build GRCh38
3417278 c.216C>T p.F72F Chr.12: 93678990-93678990 on Build GRCh38
6537790 c.124C>G p.Q42E Chr.12: 93678898-93678898 on Build GRCh38
4977853 c.139C>G p.Q47E Chr.12: 93678913-93678913 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762344890 Chr.12: 93678957-93678957 on Build GRCh38
rs760621522 Chr.12: 93678928-93678928 on Build GRCh38
rs777957131 Chr.12: 93678991-93678991 on Build GRCh38
rs375645824 Chr.12: 93679046-93679046 on Build GRCh38
rs768737669 Chr.12: 93678929-93678929 on Build GRCh38
rs988904289 Chr.12: 93678976-93678976 on Build GRCh38
rs751057807 Chr.12: 93678968-93678968 on Build GRCh38
rs758014894 Chr.12: 93678969-93678970 on Build GRCh38
rs769946054 Chr.12: 93678917-93678917 on Build GRCh38
rs777008946 Chr.12: 93678918-93678918 on Build GRCh38
rs776780462 Chr.12: 93679038-93679038 on Build GRCh38
rs755425929 Chr.12: 93678973-93678973 on Build GRCh38
rs903095279 Chr.12: 93678954-93678954 on Build GRCh38
rs550572095 Chr.12: 93679054-93679054 on Build GRCh38
rs749323581 Chr.12: 93678922-93678922 on Build GRCh38
rs764979733 Chr.12: 93678942-93678942 on Build GRCh38
rs762622157 Chr.12: 93679057-93679057 on Build GRCh38
rs765951221 Chr.12: 93678963-93678963 on Build GRCh38
rs539340280 Chr.12: 93679071-93679071 on Build GRCh38
rs768005868 Chr.12: 93679063-93679063 on Build GRCh38
rs768651771 Chr.12: 93679035-93679035 on Build GRCh38
rs768024377 Chr.12: 93678978-93678978 on Build GRCh38
rs771092659 Chr.12: 93678923-93678923 on Build GRCh38
rs749565648 Chr.12: 93678993-93678993 on Build GRCh38
rs752993084 Chr.12: 93678981-93678981 on Build GRCh38
rs190684170 Chr.12: 93679040-93679040 on Build GRCh38
rs777973744 Chr.12: 93678921-93678921 on Build GRCh38
rs1011623609 Chr.12: 93678940-93678942 on Build GRCh38
rs757453858 Chr.12: 93678995-93678995 on Build GRCh38
rs949618335 Chr.12: 93679075-93679075 on Build GRCh38
rs376616387 Chr.12: 93678983-93678983 on Build GRCh38
rs776513664 Chr.12: 93678935-93678935 on Build GRCh38
rs761709836 Chr.12: 93678939-93678939 on Build GRCh38
rs745949633 Chr.12: 93679025-93679025 on Build GRCh38
rs759051210 Chr.12: 93679060-93679060 on Build GRCh38
rs4332566 Chr.12: 93678925-93678925 on Build GRCh38
rs772979282 Chr.12: 93678945-93678945 on Build GRCh38
rs369821621 Chr.12: 93679003-93679003 on Build GRCh38
rs570406056 Chr.12: 93679059-93679059 on Build GRCh38
rs753259148 Chr.12: 93679070-93679070 on Build GRCh38

View Full Product Details