Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 19418561-19418834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSGALNACT1
Entrez Gene ID
55790
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 1
Gene Aliases
CSGalNAcT-1, ChGn, beta4GalNAcT
UniGene
Hs.613729
Gene Chromosome Location
Chr.8: 19404161-19757940 on Build GRCh38
Transcripts
NM_001130518.1 NM_018371.4 NR_024040.1 XM_006716358.2 XM_006716359.2 XM_006716360.2 XM_006716361.2 XM_006716362.2 XM_006716363.1 XM_006716364.3 XM_011544578.1 XM_011544579.1 XM_011544580.1 XM_011544582.1 XM_011544583.1 XM_011544584.1 XM_011544585.2 XM_017013623.1 XM_017013624.1 XM_017013625.1 XM_017013626.1 XM_017013627.1 XM_017013628.1 XM_017013629.1 XM_017013630.1 XM_017013631.1 XM_017013632.1 XM_017013633.1 XM_017013634.1 XM_017013635.1 XM_017013636.1 XM_017013637.1 XM_017013638.1 XM_017013639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1202384 c.1140G>T p.K380N Chr.8: 19418743-19418743 on Build GRCh38
3647049 c.1211C>T p.P404L Chr.8: 19418672-19418672 on Build GRCh38
1097808 c.1220A>G p.Q407R Chr.8: 19418663-19418663 on Build GRCh38
3647050 c.1195C>T p.H399Y Chr.8: 19418688-19418688 on Build GRCh38
5434217 c.1216G>C p.E406Q Chr.8: 19418667-19418667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913439048 Chr.8: 19418663-19418663 on Build GRCh38
rs748164265 Chr.8: 19418706-19418706 on Build GRCh38
rs553239139 Chr.8: 19418769-19418769 on Build GRCh38
rs751243403 Chr.8: 19418788-19418788 on Build GRCh38
rs539002267 Chr.8: 19418766-19418766 on Build GRCh38
rs746891603 Chr.8: 19418637-19418637 on Build GRCh38
rs775211782 Chr.8: 19418680-19418680 on Build GRCh38
rs148049722 Chr.8: 19418727-19418727 on Build GRCh38
rs750278766 Chr.8: 19418686-19418686 on Build GRCh38
rs776230720 Chr.8: 19418758-19418758 on Build GRCh38
rs374657761 Chr.8: 19418613-19418613 on Build GRCh38
rs150541306 Chr.8: 19418677-19418677 on Build GRCh38
rs536632167 Chr.8: 19418647-19418647 on Build GRCh38
rs763253247 Chr.8: 19418664-19418664 on Build GRCh38
rs927505017 Chr.8: 19418630-19418630 on Build GRCh38
rs919924412 Chr.8: 19418590-19418590 on Build GRCh38
rs375164938 Chr.8: 19418801-19418801 on Build GRCh38
rs370884744 Chr.8: 19418640-19418640 on Build GRCh38
rs773050188 Chr.8: 19418779-19418779 on Build GRCh38
rs780005169 Chr.8: 19418629-19418629 on Build GRCh38
rs762678424 Chr.8: 19418786-19418786 on Build GRCh38
rs202092533 Chr.8: 19418684-19418684 on Build GRCh38
rs769932033 Chr.8: 19418658-19418658 on Build GRCh38
rs767089032 Chr.8: 19418699-19418699 on Build GRCh38
rs745657733 Chr.8: 19418687-19418689 on Build GRCh38
rs779694868 Chr.8: 19418693-19418693 on Build GRCh38
rs756965996 Chr.8: 19418611-19418611 on Build GRCh38
rs1029253944 Chr.8: 19418635-19418635 on Build GRCh38
rs949228850 Chr.8: 19418799-19418799 on Build GRCh38
rs751570474 Chr.8: 19418694-19418694 on Build GRCh38
rs771457349 Chr.8: 19418672-19418672 on Build GRCh38
rs750189063 Chr.8: 19418620-19418620 on Build GRCh38
rs772401533 Chr.8: 19418737-19418737 on Build GRCh38
rs753221269 Chr.8: 19418683-19418683 on Build GRCh38
rs372053910 Chr.8: 19418720-19418720 on Build GRCh38
rs754951670 Chr.8: 19418793-19418793 on Build GRCh38
rs376456531 Chr.8: 19418607-19418607 on Build GRCh38
rs751096604 Chr.8: 19418591-19418591 on Build GRCh38
rs759949886 Chr.8: 19418679-19418679 on Build GRCh38
rs769374941 Chr.8: 19418779-19418781 on Build GRCh38
rs754863716 Chr.8: 19418695-19418695 on Build GRCh38
rs771386737 Chr.8: 19418729-19418729 on Build GRCh38
rs147015886 Chr.8: 19418692-19418692 on Build GRCh38
rs376108807 Chr.8: 19418628-19418628 on Build GRCh38
rs774576712 Chr.8: 19418731-19418731 on Build GRCh38
rs763729182 Chr.8: 19418682-19418682 on Build GRCh38
rs1054937925 Chr.8: 19418781-19418781 on Build GRCh38
rs756351855 Chr.8: 19418709-19418709 on Build GRCh38
rs761370634 Chr.8: 19418765-19418765 on Build GRCh38
rs773579464 Chr.8: 19418659-19418659 on Build GRCh38
rs746391651 Chr.8: 19418732-19418732 on Build GRCh38
rs576323297 Chr.8: 19418689-19418689 on Build GRCh38

View Full Product Details