Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18458146-18458393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELL
Entrez Gene ID
8178
Gene Name
elongation factor for RNA polymerase II
Gene Aliases
C19orf17, ELL1, MEN, PPP1R68
UniGene
Hs.515260
Gene Chromosome Location
Chr.19: 18442663-18522129 on Build GRCh38
Transcripts
NM_006532.3 XM_011528330.2 XM_017027335.1 XM_017027336.1 XM_017027337.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3100096 c.745-4G>A p.? Chr.19: 18458333-18458333 on Build GRCh38
6369517 c.808C>T p.Q270* Chr.19: 18458266-18458266 on Build GRCh38
3378661 c.745-3C>A p.? Chr.19: 18458332-18458332 on Build GRCh38
4075687 c.869G>A p.R290Q Chr.19: 18458205-18458205 on Build GRCh38
1205198 c.760G>A p.A254T Chr.19: 18458314-18458314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749268169 Chr.19: 18458215-18458215 on Build GRCh38
rs138119865 Chr.19: 18458288-18458288 on Build GRCh38
rs754413214 Chr.19: 18458168-18458168 on Build GRCh38
rs113111112 Chr.19: 18458343-18458343 on Build GRCh38
rs747916493 Chr.19: 18458322-18458322 on Build GRCh38
rs190373181 Chr.19: 18458206-18458206 on Build GRCh38
rs764380715 Chr.19: 18458299-18458299 on Build GRCh38
rs781267249 Chr.19: 18458348-18458350 on Build GRCh38
rs993473800 Chr.19: 18458217-18458217 on Build GRCh38
rs921229289 Chr.19: 18458318-18458318 on Build GRCh38
rs150537247 Chr.19: 18458309-18458309 on Build GRCh38
rs544681955 Chr.19: 18458258-18458258 on Build GRCh38
rs781344433 Chr.19: 18458305-18458305 on Build GRCh38
rs575679241 Chr.19: 18458209-18458209 on Build GRCh38
rs772904308 Chr.19: 18458244-18458244 on Build GRCh38
rs758685935 Chr.19: 18458197-18458197 on Build GRCh38
rs1026324748 Chr.19: 18458281-18458281 on Build GRCh38
rs370381851 Chr.19: 18458210-18458210 on Build GRCh38
rs775897838 Chr.19: 18458368-18458368 on Build GRCh38
rs776956266 Chr.19: 18458270-18458270 on Build GRCh38
rs139727451 Chr.19: 18458243-18458243 on Build GRCh38
rs770292588 Chr.19: 18458344-18458344 on Build GRCh38
rs779813825 Chr.19: 18458212-18458212 on Build GRCh38
rs770309503 Chr.19: 18458361-18458361 on Build GRCh38
rs778775562 Chr.19: 18458319-18458319 on Build GRCh38
rs750224144 Chr.19: 18458368-18458371 on Build GRCh38
rs145570868 Chr.19: 18458274-18458274 on Build GRCh38
rs199822246 Chr.19: 18458194-18458194 on Build GRCh38
rs765728277 Chr.19: 18458278-18458278 on Build GRCh38
rs374724148 Chr.19: 18458342-18458342 on Build GRCh38
rs751861329 Chr.19: 18458300-18458300 on Build GRCh38
rs757361492 Chr.19: 18458301-18458301 on Build GRCh38
rs761853013 Chr.19: 18458334-18458334 on Build GRCh38
rs758234072 Chr.19: 18458340-18458340 on Build GRCh38
rs368978110 Chr.19: 18458363-18458363 on Build GRCh38
rs748004787 Chr.19: 18458236-18458236 on Build GRCh38
rs778774419 Chr.19: 18458231-18458231 on Build GRCh38
rs148864996 Chr.19: 18458196-18458196 on Build GRCh38
rs776011018 Chr.19: 18458306-18458306 on Build GRCh38
rs889702377 Chr.19: 18458173-18458173 on Build GRCh38
rs753088902 Chr.19: 18458285-18458285 on Build GRCh38
rs759782160 Chr.19: 18458191-18458191 on Build GRCh38
rs554368355 Chr.19: 18458167-18458167 on Build GRCh38
rs112087517 Chr.19: 18458279-18458279 on Build GRCh38
rs561148008 Chr.19: 18458350-18458350 on Build GRCh38
rs760150831 Chr.19: 18458246-18458246 on Build GRCh38

View Full Product Details