Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 33524843-33525086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD2
Entrez Gene ID
114784
Gene Name
CUB and Sushi multiple domains 2
Gene Aliases
dJ1007G16.1, dJ1007G16.2, dJ947L8.1
UniGene
Hs.656915
Gene Chromosome Location
Chr.1: 33513999-34165842 on Build GRCh38
Transcripts
NM_001281956.1 NM_052896.4 XM_017000185.1 XM_017000186.1 XM_017000187.1 XM_017000188.1 XM_017000189.1 XM_017000190.1 XM_017000194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1321140 c.10377C>T p.G3459G Chr.1: 33524901-33524901 on Build GRCh38
426124 c.9893G>A p.G3298D Chr.1: 33524953-33524953 on Build GRCh38
5906622 c.9836G>T p.W3279L Chr.1: 33525010-33525010 on Build GRCh38
160244 c.9933C>T p.I3311I Chr.1: 33524913-33524913 on Build GRCh38
7213871 c.10371C>A p.H3457Q Chr.1: 33524907-33524907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754453331 Chr.1: 33524942-33524942 on Build GRCh38
rs757782719 Chr.1: 33524953-33524953 on Build GRCh38
rs771646055 Chr.1: 33524970-33524970 on Build GRCh38
rs966936325 Chr.1: 33525053-33525053 on Build GRCh38
rs148333754 Chr.1: 33524986-33524986 on Build GRCh38
rs192427685 Chr.1: 33525035-33525035 on Build GRCh38
rs897008235 Chr.1: 33524921-33524921 on Build GRCh38
rs759708482 Chr.1: 33525057-33525057 on Build GRCh38
rs1020441283 Chr.1: 33525040-33525040 on Build GRCh38
rs752839094 Chr.1: 33524913-33524913 on Build GRCh38
rs201348886 Chr.1: 33525065-33525065 on Build GRCh38
rs761740866 Chr.1: 33525012-33525012 on Build GRCh38
rs149550450 Chr.1: 33524917-33524917 on Build GRCh38
rs373776858 Chr.1: 33524876-33524876 on Build GRCh38
rs888925923 Chr.1: 33524936-33524936 on Build GRCh38
rs776593570 Chr.1: 33524869-33524869 on Build GRCh38
rs761738854 Chr.1: 33524885-33524885 on Build GRCh38
rs199837901 Chr.1: 33524868-33524868 on Build GRCh38
rs139653382 Chr.1: 33525038-33525038 on Build GRCh38
rs769727125 Chr.1: 33525024-33525024 on Build GRCh38
rs377744817 Chr.1: 33524877-33524877 on Build GRCh38
rs755183571 Chr.1: 33524901-33524901 on Build GRCh38
rs933221905 Chr.1: 33524895-33524895 on Build GRCh38
rs1024266803 Chr.1: 33524981-33524981 on Build GRCh38
rs767726610 Chr.1: 33524912-33524912 on Build GRCh38
rs778291440 Chr.1: 33524960-33524960 on Build GRCh38
rs1001211833 Chr.1: 33525002-33525002 on Build GRCh38
rs187099966 Chr.1: 33524900-33524900 on Build GRCh38
rs374791495 Chr.1: 33524966-33524966 on Build GRCh38
rs768509269 Chr.1: 33524995-33524995 on Build GRCh38
rs779808938 Chr.1: 33524971-33524971 on Build GRCh38
rs767461458 Chr.1: 33525061-33525061 on Build GRCh38
rs994400307 Chr.1: 33524931-33524931 on Build GRCh38
rs766378569 Chr.1: 33524893-33524893 on Build GRCh38
rs776401540 Chr.1: 33524998-33524998 on Build GRCh38

View Full Product Details