Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109924037-109924306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF1
Entrez Gene ID
1435
Gene Name
colony stimulating factor 1
Gene Aliases
CSF-1, MCSF
UniGene
Hs.173894
Gene Chromosome Location
Chr.1: 109910506-109930994 on Build GRCh38
Transcripts
NM_000757.5 NM_172210.2 NM_172211.3 NM_172212.2 XM_017000369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5352821 c.1430C>A p.T477N Chr.1: 109924051-109924051 on Build GRCh38
7114580 c.1443T>C p.H481H Chr.1: 109924064-109924064 on Build GRCh38
3471014 c.1538G>A p.G513E Chr.1: 109924159-109924159 on Build GRCh38
5590374 c.1424C>T p.P475L Chr.1: 109924045-109924045 on Build GRCh38
4172112 c.1529T>A p.L510Q Chr.1: 109924150-109924150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750931110 Chr.1: 109924208-109924208 on Build GRCh38
rs556461036 Chr.1: 109924178-109924178 on Build GRCh38
rs2229166 Chr.1: 109924188-109924188 on Build GRCh38
rs749554501 Chr.1: 109924063-109924063 on Build GRCh38
rs886224538 Chr.1: 109924249-109924249 on Build GRCh38
rs750993174 Chr.1: 109924151-109924151 on Build GRCh38
rs755086255 Chr.1: 109924221-109924221 on Build GRCh38
rs1014792253 Chr.1: 109924250-109924250 on Build GRCh38
rs34463755 Chr.1: 109924230-109924230 on Build GRCh38
rs762038446 Chr.1: 109924205-109924205 on Build GRCh38
rs373422212 Chr.1: 109924105-109924105 on Build GRCh38
rs767858113 Chr.1: 109924148-109924148 on Build GRCh38
rs760118409 Chr.1: 109924093-109924093 on Build GRCh38
rs945181969 Chr.1: 109924081-109924081 on Build GRCh38
rs12721516 Chr.1: 109924108-109924108 on Build GRCh38
rs1004336649 Chr.1: 109924261-109924262 on Build GRCh38
rs767959767 Chr.1: 109924207-109924207 on Build GRCh38
rs771970619 Chr.1: 109924077-109924077 on Build GRCh38
rs770406825 Chr.1: 109924186-109924186 on Build GRCh38
rs768537404 Chr.1: 109924072-109924072 on Build GRCh38
rs370727907 Chr.1: 109924262-109924262 on Build GRCh38
rs755209927 Chr.1: 109924162-109924162 on Build GRCh38
rs140931502 Chr.1: 109924155-109924155 on Build GRCh38
rs149758130 Chr.1: 109924263-109924263 on Build GRCh38
rs140090023 Chr.1: 109924112-109924112 on Build GRCh38
rs576127753 Chr.1: 109924185-109924185 on Build GRCh38
rs879861630 Chr.1: 109924254-109924254 on Build GRCh38
rs778943546 Chr.1: 109924222-109924222 on Build GRCh38
rs761311283 Chr.1: 109924210-109924210 on Build GRCh38
rs3208449 Chr.1: 109924154-109924154 on Build GRCh38
rs778618530 Chr.1: 109924076-109924076 on Build GRCh38
rs564534457 Chr.1: 109924281-109924281 on Build GRCh38
rs333971 Chr.1: 109924087-109924087 on Build GRCh38
rs775226242 Chr.1: 109924191-109924191 on Build GRCh38
rs201860266 Chr.1: 109924137-109924137 on Build GRCh38
rs541078222 Chr.1: 109924280-109924280 on Build GRCh38
rs367579394 Chr.1: 109924094-109924094 on Build GRCh38
rs778760181 Chr.1: 109924172-109924172 on Build GRCh38
rs769264432 Chr.1: 109924189-109924189 on Build GRCh38
rs759237248 Chr.1: 109924103-109924103 on Build GRCh38
rs766551433 Chr.1: 109924214-109924214 on Build GRCh38
rs147216763 Chr.1: 109924157-109924157 on Build GRCh38
rs367810859 Chr.1: 109924218-109924218 on Build GRCh38
rs140567876 Chr.1: 109924184-109924184 on Build GRCh38

View Full Product Details