Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23509953-23510178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F2
Entrez Gene ID
1870
Gene Name
E2F transcription factor 2
Gene Aliases
E2F-2
UniGene
Hs.194333
Gene Chromosome Location
Chr.1: 23505696-23531266 on Build GRCh38
Transcripts
NM_004091.3 XM_005245748.3 XM_005245749.3 XM_011540868.2 XM_011540870.2 XM_011540871.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1320337 c.1121T>A p.L374Q Chr.1: 23510073-23510073 on Build GRCh38
82118 c.1239G>T p.L413L Chr.1: 23509955-23509955 on Build GRCh38
906301 c.1156G>A p.E386K Chr.1: 23510038-23510038 on Build GRCh38
2049175 c.1058C>T p.A353V Chr.1: 23510136-23510136 on Build GRCh38
6329732 c.1188C>T p.C396C Chr.1: 23510006-23510006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761768883 Chr.1: 23510074-23510074 on Build GRCh38
rs142571891 Chr.1: 23509986-23509986 on Build GRCh38
rs1014160601 Chr.1: 23509981-23509981 on Build GRCh38
rs369246681 Chr.1: 23510081-23510081 on Build GRCh38
rs926264748 Chr.1: 23510055-23510055 on Build GRCh38
rs763913178 Chr.1: 23510115-23510115 on Build GRCh38
rs767232696 Chr.1: 23510111-23510111 on Build GRCh38
rs530082478 Chr.1: 23510061-23510061 on Build GRCh38
rs777829086 Chr.1: 23510017-23510017 on Build GRCh38
rs745450429 Chr.1: 23510136-23510136 on Build GRCh38
rs773372814 Chr.1: 23510099-23510099 on Build GRCh38
rs754726067 Chr.1: 23509979-23509979 on Build GRCh38
rs755992777 Chr.1: 23510113-23510113 on Build GRCh38
rs756781681 Chr.1: 23510129-23510129 on Build GRCh38
rs752620979 Chr.1: 23510009-23510009 on Build GRCh38
rs548500891 Chr.1: 23510018-23510018 on Build GRCh38
rs886819893 Chr.1: 23510010-23510010 on Build GRCh38
rs778672424 Chr.1: 23510022-23510022 on Build GRCh38
rs772511958 Chr.1: 23510059-23510059 on Build GRCh38
rs151287835 Chr.1: 23510060-23510060 on Build GRCh38
rs1049336725 Chr.1: 23510125-23510125 on Build GRCh38
rs752426258 Chr.1: 23510112-23510112 on Build GRCh38
rs765424043 Chr.1: 23509976-23509976 on Build GRCh38
rs370898219 Chr.1: 23510013-23510013 on Build GRCh38
rs750682155 Chr.1: 23509977-23509977 on Build GRCh38
rs777245600 Chr.1: 23510066-23510066 on Build GRCh38
rs763176248 Chr.1: 23510109-23510109 on Build GRCh38
rs778584443 Chr.1: 23510135-23510135 on Build GRCh38
rs866938398 Chr.1: 23510094-23510094 on Build GRCh38
rs753848134 Chr.1: 23510127-23510127 on Build GRCh38
rs183643474 Chr.1: 23510050-23510050 on Build GRCh38
rs1016525076 Chr.1: 23510025-23510025 on Build GRCh38

View Full Product Details