Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 100152112-100152381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERLIN1
Entrez Gene ID
10613
Gene Name
ER lipid raft associated 1
Gene Aliases
C10orf69, Erlin-1, KE04, KEO4, SPFH1, SPG62
UniGene
Hs.150087
Gene Chromosome Location
Chr.10: 100150090-100186057 on Build GRCh38
Transcripts
NM_001100626.1 NM_006459.3 XM_005269442.3 XM_017015541.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3751487 c.871A>G p.I291V Chr.10: 100152307-100152307 on Build GRCh38
3709740 c.1018G>A p.V340I Chr.10: 100152160-100152160 on Build GRCh38
1345268 c.952G>A p.D318N Chr.10: 100152226-100152226 on Build GRCh38
6325309 c.898G>T p.G300C Chr.10: 100152280-100152280 on Build GRCh38
3665437 c.890T>G p.I297S Chr.10: 100152288-100152288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769511375 Chr.10: 100152194-100152194 on Build GRCh38
rs778506214 Chr.10: 100152354-100152354 on Build GRCh38
rs768523536 Chr.10: 100152211-100152211 on Build GRCh38
rs766711343 Chr.10: 100152249-100152249 on Build GRCh38
rs745419107 Chr.10: 100152297-100152297 on Build GRCh38
rs775229931 Chr.10: 100152197-100152197 on Build GRCh38
rs929178959 Chr.10: 100152313-100152313 on Build GRCh38
rs756772947 Chr.10: 100152285-100152285 on Build GRCh38
rs771868689 Chr.10: 100152302-100152302 on Build GRCh38
rs147388040 Chr.10: 100152244-100152244 on Build GRCh38
rs756477696 Chr.10: 100152150-100152150 on Build GRCh38
rs916645978 Chr.10: 100152233-100152233 on Build GRCh38
rs199928340 Chr.10: 100152163-100152163 on Build GRCh38
rs1017693350 Chr.10: 100152323-100152323 on Build GRCh38
rs766692560 Chr.10: 100152348-100152348 on Build GRCh38
rs774243081 Chr.10: 100152322-100152322 on Build GRCh38
rs879856876 Chr.10: 100152240-100152240 on Build GRCh38
rs768507729 Chr.10: 100152315-100152315 on Build GRCh38
rs746815062 Chr.10: 100152167-100152168 on Build GRCh38
rs781032219 Chr.10: 100152359-100152359 on Build GRCh38
rs773362257 Chr.10: 100152236-100152236 on Build GRCh38
rs765743611 Chr.10: 100152358-100152358 on Build GRCh38
rs369725736 Chr.10: 100152141-100152141 on Build GRCh38
rs780862850 Chr.10: 100152192-100152192 on Build GRCh38
rs777934587 Chr.10: 100152141-100152142 on Build GRCh38
rs772075743 Chr.10: 100152340-100152340 on Build GRCh38
rs2862954 Chr.10: 100152307-100152307 on Build GRCh38
rs374471439 Chr.10: 100152275-100152275 on Build GRCh38
rs887722937 Chr.10: 100152321-100152321 on Build GRCh38
rs560433756 Chr.10: 100152214-100152214 on Build GRCh38
rs765831888 Chr.10: 100152267-100152267 on Build GRCh38
rs780521035 Chr.10: 100152294-100152294 on Build GRCh38
rs779695614 Chr.10: 100152304-100152304 on Build GRCh38
rs777056627 Chr.10: 100152352-100152352 on Build GRCh38
rs779572900 Chr.10: 100152161-100152161 on Build GRCh38
rs371568828 Chr.10: 100152256-100152256 on Build GRCh38
rs756947978 Chr.10: 100152350-100152350 on Build GRCh38
rs767541138 Chr.10: 100152223-100152223 on Build GRCh38
rs373292585 Chr.10: 100152160-100152160 on Build GRCh38
rs377025416 Chr.10: 100152212-100152212 on Build GRCh38
rs976456038 Chr.10: 100152272-100152272 on Build GRCh38
rs373603837 Chr.10: 100152152-100152152 on Build GRCh38
rs552222409 Chr.10: 100152342-100152342 on Build GRCh38
rs375721397 Chr.10: 100152346-100152346 on Build GRCh38
rs755659573 Chr.10: 100152300-100152300 on Build GRCh38
rs747985150 Chr.10: 100152338-100152338 on Build GRCh38
rs755447558 Chr.10: 100152262-100152262 on Build GRCh38
rs900216990 Chr.10: 100152339-100152339 on Build GRCh38
rs749089399 Chr.10: 100152198-100152198 on Build GRCh38
rs747951974 Chr.10: 100152357-100152357 on Build GRCh38
rs949502207 Chr.10: 100152287-100152287 on Build GRCh38

View Full Product Details