Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140671524-140671779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DND1
Entrez Gene ID
373863
Gene Name
DND microRNA-mediated repression inhibitor 1
Gene Aliases
RBMS4
UniGene
Hs.591262
Gene Chromosome Location
Chr.5: 140670796-140673586 on Build GRCh38
Transcripts
NM_194249.2 XM_005268469.2 XM_017009600.1

Gene Symbol
WDR55
Entrez Gene ID
54853
Gene Name
WD repeat domain 55
Gene Aliases
-
UniGene
Hs.286261
Gene Chromosome Location
Chr.5: 140664799-140674119 on Build GRCh38
Transcripts
NM_194249.2 XM_005268469.2 XM_017009600.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1061744 c.764C>T p.A255V Chr.5: 140671591-140671591 on Build GRCh38
5709477 c.637G>C p.V213L Chr.5: 140671718-140671718 on Build GRCh38
5076432 c.812_813delCT p.P271fs*47 Chr.5: 140671542-140671543 on Build GRCh38
1061745 c.673C>T p.R225C Chr.5: 140671682-140671682 on Build GRCh38
1619578 c.647T>A p.L216H Chr.5: 140671708-140671708 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003596439 Chr.5: 140671713-140671713 on Build GRCh38
rs527677101 Chr.5: 140671608-140671608 on Build GRCh38
rs753368143 Chr.5: 140671640-140671640 on Build GRCh38
rs763672102 Chr.5: 140671632-140671632 on Build GRCh38
rs756967253 Chr.5: 140671647-140671647 on Build GRCh38
rs185834706 Chr.5: 140671589-140671589 on Build GRCh38
rs770100890 Chr.5: 140671682-140671682 on Build GRCh38
rs778524394 Chr.5: 140671650-140671650 on Build GRCh38
rs773644539 Chr.5: 140671561-140671561 on Build GRCh38
rs774939347 Chr.5: 140671705-140671705 on Build GRCh38
rs533708303 Chr.5: 140671652-140671652 on Build GRCh38
rs941678913 Chr.5: 140671569-140671569 on Build GRCh38
rs755742033 Chr.5: 140671611-140671611 on Build GRCh38
rs569230593 Chr.5: 140671639-140671639 on Build GRCh38
rs767033035 Chr.5: 140671597-140671597 on Build GRCh38
rs747080694 Chr.5: 140671655-140671655 on Build GRCh38
rs148957130 Chr.5: 140671663-140671663 on Build GRCh38
rs763474206 Chr.5: 140671735-140671735 on Build GRCh38
rs773413092 Chr.5: 140671686-140671686 on Build GRCh38
rs1052037563 Chr.5: 140671618-140671618 on Build GRCh38
rs758305425 Chr.5: 140671653-140671653 on Build GRCh38
rs753049676 Chr.5: 140671628-140671628 on Build GRCh38
rs771550759 Chr.5: 140671696-140671696 on Build GRCh38
rs1042383882 Chr.5: 140671700-140671700 on Build GRCh38
rs781360938 Chr.5: 140671680-140671680 on Build GRCh38
rs377680919 Chr.5: 140671563-140671563 on Build GRCh38
rs143703434 Chr.5: 140671681-140671681 on Build GRCh38
rs145678669 Chr.5: 140671654-140671654 on Build GRCh38
rs141336765 Chr.5: 140671719-140671719 on Build GRCh38
rs149777619 Chr.5: 140671605-140671605 on Build GRCh38
rs867899860 Chr.5: 140671697-140671697 on Build GRCh38
rs146619681 Chr.5: 140671687-140671687 on Build GRCh38
rs902596965 Chr.5: 140671709-140671709 on Build GRCh38

View Full Product Details