Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24575192-24575465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSG
Entrez Gene ID
1511
Gene Name
cathepsin G
Gene Aliases
CATG, CG
UniGene
Hs.421724
Gene Chromosome Location
Chr.14: 24573518-24576260 on Build GRCh38
Transcripts
NM_001911.2 XM_011536499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1369424 c.155T>G p.F52C Chr.14: 24575313-24575313 on Build GRCh38
4513193 c.92C>A p.S31Y Chr.14: 24575376-24575376 on Build GRCh38
4577818 c.66C>A p.I22I Chr.14: 24575402-24575402 on Build GRCh38
6227383 c.170A>T p.D57V Chr.14: 24575298-24575298 on Build GRCh38
955106 c.107C>T p.A36V Chr.14: 24575361-24575361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374772459 Chr.14: 24575412-24575412 on Build GRCh38
rs916877861 Chr.14: 24575374-24575374 on Build GRCh38
rs199634129 Chr.14: 24575410-24575410 on Build GRCh38
rs138881085 Chr.14: 24575218-24575218 on Build GRCh38
rs756467866 Chr.14: 24575248-24575248 on Build GRCh38
rs751585793 Chr.14: 24575408-24575408 on Build GRCh38
rs922075961 Chr.14: 24575404-24575404 on Build GRCh38
rs761109514 Chr.14: 24575223-24575223 on Build GRCh38
rs762863588 Chr.14: 24575389-24575389 on Build GRCh38
rs748224511 Chr.14: 24575251-24575251 on Build GRCh38
rs112417622 Chr.14: 24575383-24575383 on Build GRCh38
rs776029478 Chr.14: 24575346-24575346 on Build GRCh38
rs756051936 Chr.14: 24575441-24575441 on Build GRCh38
rs367993614 Chr.14: 24575423-24575423 on Build GRCh38
rs974936619 Chr.14: 24575371-24575371 on Build GRCh38
rs529487105 Chr.14: 24575394-24575394 on Build GRCh38
rs765016127 Chr.14: 24575367-24575367 on Build GRCh38
rs780991621 Chr.14: 24575228-24575229 on Build GRCh38
rs764964221 Chr.14: 24575241-24575241 on Build GRCh38
rs779257437 Chr.14: 24575431-24575431 on Build GRCh38
rs377162733 Chr.14: 24575360-24575360 on Build GRCh38
rs750317988 Chr.14: 24575244-24575244 on Build GRCh38
rs549168777 Chr.14: 24575245-24575246 on Build GRCh38
rs779154527 Chr.14: 24575328-24575328 on Build GRCh38
rs752608186 Chr.14: 24575287-24575287 on Build GRCh38
rs746315813 Chr.14: 24575329-24575329 on Build GRCh38
rs750179849 Chr.14: 24575373-24575373 on Build GRCh38
rs756309618 Chr.14: 24575256-24575256 on Build GRCh38
rs149063733 Chr.14: 24575365-24575365 on Build GRCh38
rs922808647 Chr.14: 24575320-24575320 on Build GRCh38
rs886590242 Chr.14: 24575411-24575411 on Build GRCh38
rs146445852 Chr.14: 24575344-24575344 on Build GRCh38
rs564479432 Chr.14: 24575217-24575217 on Build GRCh38
rs907879784 Chr.14: 24575445-24575445 on Build GRCh38
rs749550003 Chr.14: 24575299-24575299 on Build GRCh38
rs200884114 Chr.14: 24575237-24575237 on Build GRCh38
rs201990007 Chr.14: 24575361-24575361 on Build GRCh38
rs771274266 Chr.14: 24575309-24575309 on Build GRCh38
rs184648249 Chr.14: 24575440-24575440 on Build GRCh38
rs112407280 Chr.14: 24575358-24575358 on Build GRCh38
rs930874791 Chr.14: 24575387-24575387 on Build GRCh38
rs372731166 Chr.14: 24575297-24575297 on Build GRCh38
rs1000333362 Chr.14: 24575228-24575228 on Build GRCh38
rs756219791 Chr.14: 24575409-24575409 on Build GRCh38
rs764462214 Chr.14: 24575225-24575225 on Build GRCh38
rs187232086 Chr.14: 24575230-24575230 on Build GRCh38

View Full Product Details