Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107903391-107903634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLD
Entrez Gene ID
1738
Gene Name
dihydrolipoamide dehydrogenase
Gene Aliases
DLDD, DLDH, E3, GCSL, LAD, PHE3
UniGene
Hs.131711
Gene Chromosome Location
Chr.7: 107891107-107921198 on Build GRCh38
Transcripts
NM_000108.4 NM_001289750.1 NM_001289751.1 NM_001289752.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1083922 c.313G>T p.D105Y Chr.7: 107903523-107903523 on Build GRCh38
3255004 c.320C>T p.A107V Chr.7: 107903530-107903530 on Build GRCh38
5560223 c.298A>G p.M100V Chr.7: 107903508-107903508 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371072449 Chr.7: 107903554-107903554 on Build GRCh38
rs754528609 Chr.7: 107903574-107903574 on Build GRCh38
rs992452328 Chr.7: 107903458-107903458 on Build GRCh38
rs749777272 Chr.7: 107903591-107903591 on Build GRCh38
rs765105905 Chr.7: 107903529-107903529 on Build GRCh38
rs893865374 Chr.7: 107903515-107903515 on Build GRCh38
rs916946250 Chr.7: 107903466-107903466 on Build GRCh38
rs761564212 Chr.7: 107903495-107903495 on Build GRCh38
rs796051950 Chr.7: 107903516-107903516 on Build GRCh38
rs775551311 Chr.7: 107903435-107903435 on Build GRCh38
rs1041605197 Chr.7: 107903509-107903509 on Build GRCh38
rs766075550 Chr.7: 107903550-107903550 on Build GRCh38
rs1130477 Chr.7: 107903521-107903521 on Build GRCh38
rs374458520 Chr.7: 107903579-107903579 on Build GRCh38
rs746121954 Chr.7: 107903429-107903429 on Build GRCh38
rs373587265 Chr.7: 107903459-107903459 on Build GRCh38
rs760731186 Chr.7: 107903450-107903450 on Build GRCh38
rs144617361 Chr.7: 107903492-107903492 on Build GRCh38
rs150502288 Chr.7: 107903431-107903431 on Build GRCh38
rs752671238 Chr.7: 107903557-107903557 on Build GRCh38
rs751938025 Chr.7: 107903446-107903446 on Build GRCh38
rs758078257 Chr.7: 107903532-107903532 on Build GRCh38
rs11538953 Chr.7: 107903505-107903505 on Build GRCh38
rs138398782 Chr.7: 107903531-107903531 on Build GRCh38
rs376524199 Chr.7: 107903603-107903620 on Build GRCh38
rs751168608 Chr.7: 107903566-107903566 on Build GRCh38
rs945939975 Chr.7: 107903507-107903507 on Build GRCh38

View Full Product Details