Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44943625-44943898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM207A
Entrez Gene ID
85395
Gene Name
family with sequence similarity 207 member A
Gene Aliases
C21orf70, PRED56
UniGene
Hs.410830
Gene Chromosome Location
Chr.21: 44939697-44976989 on Build GRCh38
Transcripts
NM_001316983.1 NM_001316984.1 NM_001316985.1 NM_001316986.1 NM_001316987.1 NM_001316988.1 NM_058190.3 XM_017028481.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4483185 c.267C>T p.V89V Chr.21: 44943821-44943821 on Build GRCh38
4483389 c.269C>T p.P90L Chr.21: 44943823-44943823 on Build GRCh38
6659316 c.244G>A p.A82T Chr.21: 44943798-44943798 on Build GRCh38
478657 c.207G>A p.L69L Chr.21: 44943761-44943761 on Build GRCh38
4877459 c.229G>A p.V77I Chr.21: 44943783-44943783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540825858 Chr.21: 44943828-44943828 on Build GRCh38
rs145890659 Chr.21: 44943742-44943742 on Build GRCh38
rs138701512 Chr.21: 44943765-44943765 on Build GRCh38
rs767058573 Chr.21: 44943869-44943869 on Build GRCh38
rs763483733 Chr.21: 44943660-44943660 on Build GRCh38
rs745578671 Chr.21: 44943802-44943802 on Build GRCh38
rs11539072 Chr.21: 44943790-44943790 on Build GRCh38
rs752147754 Chr.21: 44943778-44943822 on Build GRCh38
rs756367183 Chr.21: 44943868-44943869 on Build GRCh38
rs573541268 Chr.21: 44943822-44943822 on Build GRCh38
rs200952167 Chr.21: 44943806-44943806 on Build GRCh38
rs199972945 Chr.21: 44943872-44943872 on Build GRCh38
rs754429487 Chr.21: 44943676-44943676 on Build GRCh38
rs759567934 Chr.21: 44943850-44943850 on Build GRCh38
rs745316372 Chr.21: 44943737-44943737 on Build GRCh38
rs186057333 Chr.21: 44943740-44943740 on Build GRCh38
rs758841215 Chr.21: 44943792-44943792 on Build GRCh38
rs576306304 Chr.21: 44943783-44943783 on Build GRCh38
rs750981303 Chr.21: 44943803-44943803 on Build GRCh38
rs374466447 Chr.21: 44943782-44943782 on Build GRCh38
rs978039957 Chr.21: 44943667-44943667 on Build GRCh38
rs754268973 Chr.21: 44943778-44943778 on Build GRCh38
rs368028677 Chr.21: 44943853-44943853 on Build GRCh38
rs140146705 Chr.21: 44943819-44943819 on Build GRCh38
rs755293907 Chr.21: 44943684-44943684 on Build GRCh38
rs371091838 Chr.21: 44943777-44943777 on Build GRCh38
rs559028093 Chr.21: 44943851-44943851 on Build GRCh38
rs549623335 Chr.21: 44943813-44943813 on Build GRCh38
rs773438173 Chr.21: 44943767-44943767 on Build GRCh38
rs768055719 Chr.21: 44943814-44943814 on Build GRCh38
rs766547272 Chr.21: 44943773-44943773 on Build GRCh38
rs767748108 Chr.21: 44943663-44943663 on Build GRCh38
rs781025430 Chr.21: 44943703-44943703 on Build GRCh38
rs376432825 Chr.21: 44943854-44943854 on Build GRCh38
rs760999581 Chr.21: 44943825-44943825 on Build GRCh38
rs889359431 Chr.21: 44943844-44943844 on Build GRCh38
rs770893845 Chr.21: 44943833-44943833 on Build GRCh38
rs766740509 Chr.21: 44943675-44943675 on Build GRCh38
rs780976052 Chr.21: 44943795-44943795 on Build GRCh38
rs760731574 Chr.21: 44943772-44943772 on Build GRCh38
rs1036755699 Chr.21: 44943817-44943817 on Build GRCh38
rs535219131 Chr.21: 44943686-44943686 on Build GRCh38
rs371932148 Chr.21: 44943814-44943814 on Build GRCh38
rs764673084 Chr.21: 44943650-44943652 on Build GRCh38
rs370556723 Chr.21: 44943664-44943664 on Build GRCh38
rs768595822 Chr.21: 44943809-44943809 on Build GRCh38
rs78520285 Chr.21: 44943692-44943692 on Build GRCh38
rs755561561 Chr.21: 44943779-44943779 on Build GRCh38
rs757696864 Chr.21: 44943784-44943828 on Build GRCh38
rs771913102 Chr.21: 44943818-44943818 on Build GRCh38
rs768312237 Chr.21: 44943752-44943752 on Build GRCh38
rs374980554 Chr.21: 44943837-44943837 on Build GRCh38
rs142143446 Chr.21: 44943738-44943738 on Build GRCh38
rs373722100 Chr.21: 44943672-44943672 on Build GRCh38
rs750076263 Chr.21: 44943862-44943862 on Build GRCh38
rs1021438732 Chr.21: 44943871-44943871 on Build GRCh38
rs761624834 Chr.21: 44943764-44943764 on Build GRCh38
rs926517567 Chr.21: 44943746-44943746 on Build GRCh38
rs755545736 Chr.21: 44943805-44943805 on Build GRCh38
rs567793966 Chr.21: 44943659-44943659 on Build GRCh38
rs367559441 Chr.21: 44943691-44943691 on Build GRCh38
rs988150550 Chr.21: 44943705-44943705 on Build GRCh38
rs201505042 Chr.21: 44943770-44943770 on Build GRCh38
rs750557151 Chr.21: 44943815-44943816 on Build GRCh38
rs1021050291 Chr.21: 44943725-44943725 on Build GRCh38
rs1007805524 Chr.21: 44943848-44943848 on Build GRCh38

View Full Product Details