Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 4068662-4068923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM217A
Entrez Gene ID
222826
Gene Name
family with sequence similarity 217 member A
Gene Aliases
C6orf146
UniGene
Hs.657342
Gene Chromosome Location
Chr.6: 4068352-4087344 on Build GRCh38
Transcripts
NM_173563.2 XM_005248951.3 XM_006715025.2 XM_006715026.3 XM_011514413.2 XM_011514414.2 XM_011514415.1 XM_011514416.2 XM_011514417.1 XM_017010480.1 XM_017010481.1 XM_017010482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1079132 c.1372G>T p.E458* Chr.6: 4068851-4068851 on Build GRCh38
1079131 c.1421G>A p.R474Q Chr.6: 4068802-4068802 on Build GRCh38
5870622 c.1369G>A p.E457K Chr.6: 4068854-4068854 on Build GRCh38
3430654 c.1326G>A p.M442I Chr.6: 4068897-4068897 on Build GRCh38
6529720 c.1322C>G p.P441R Chr.6: 4068901-4068901 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751042959 Chr.6: 4068709-4068709 on Build GRCh38
rs199940214 Chr.6: 4068779-4068779 on Build GRCh38
rs746127562 Chr.6: 4068723-4068723 on Build GRCh38
rs757866954 Chr.6: 4068889-4068889 on Build GRCh38
rs762547400 Chr.6: 4068786-4068786 on Build GRCh38
rs144485598 Chr.6: 4068837-4068837 on Build GRCh38
rs778497109 Chr.6: 4068749-4068752 on Build GRCh38
rs538409355 Chr.6: 4068711-4068711 on Build GRCh38
rs756714408 Chr.6: 4068710-4068710 on Build GRCh38
rs775462401 Chr.6: 4068729-4068729 on Build GRCh38
rs758570734 Chr.6: 4068824-4068824 on Build GRCh38
rs747996472 Chr.6: 4068750-4068751 on Build GRCh38
rs888040752 Chr.6: 4068804-4068804 on Build GRCh38
rs757817856 Chr.6: 4068813-4068813 on Build GRCh38
rs768594931 Chr.6: 4068735-4068735 on Build GRCh38
rs200799506 Chr.6: 4068860-4068860 on Build GRCh38
rs774218991 Chr.6: 4068867-4068867 on Build GRCh38
rs751049253 Chr.6: 4068796-4068796 on Build GRCh38
rs993942124 Chr.6: 4068857-4068857 on Build GRCh38
rs371477063 Chr.6: 4068727-4068727 on Build GRCh38
rs916660558 Chr.6: 4068752-4068752 on Build GRCh38
rs767188381 Chr.6: 4068883-4068883 on Build GRCh38
rs769733195 Chr.6: 4068849-4068849 on Build GRCh38
rs139841919 Chr.6: 4068698-4068698 on Build GRCh38
rs745800947 Chr.6: 4068740-4068740 on Build GRCh38
rs749675687 Chr.6: 4068712-4068712 on Build GRCh38
rs776480454 Chr.6: 4068747-4068747 on Build GRCh38
rs183033133 Chr.6: 4068859-4068859 on Build GRCh38
rs10485172 Chr.6: 4068899-4068899 on Build GRCh38
rs374965334 Chr.6: 4068866-4068866 on Build GRCh38
rs576082742 Chr.6: 4068723-4068728 on Build GRCh38
rs781167221 Chr.6: 4068826-4068826 on Build GRCh38
rs375222136 Chr.6: 4068878-4068878 on Build GRCh38
rs144438667 Chr.6: 4068748-4068748 on Build GRCh38
rs754475677 Chr.6: 4068802-4068802 on Build GRCh38
rs574472718 Chr.6: 4068884-4068884 on Build GRCh38
rs766010596 Chr.6: 4068793-4068793 on Build GRCh38
rs369347224 Chr.6: 4068841-4068841 on Build GRCh38
rs201828721 Chr.6: 4068847-4068847 on Build GRCh38
rs764997056 Chr.6: 4068770-4068770 on Build GRCh38
rs141691461 Chr.6: 4068716-4068716 on Build GRCh38
rs375725382 Chr.6: 4068803-4068803 on Build GRCh38
rs760696595 Chr.6: 4068734-4068734 on Build GRCh38
rs148412142 Chr.6: 4068814-4068814 on Build GRCh38
rs1011333843 Chr.6: 4068852-4068852 on Build GRCh38

View Full Product Details