Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 114522970-114523207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXPE1
Entrez Gene ID
120400
Gene Name
neurexophilin and PC-esterase domain family member 1
Gene Aliases
FAM55A
UniGene
Hs.721328
Gene Chromosome Location
Chr.11: 114518946-114560244 on Build GRCh38
Transcripts
NM_152315.3 XM_011542594.2 XM_011542595.2 XM_011542596.2 XM_011542597.2 XM_011542598.2 XM_011542599.2 XM_017017207.1 XM_017017209.1 XM_017017210.1

Gene Symbol
NXPE2
Entrez Gene ID
120406
Gene Name
neurexophilin and PC-esterase domain family member 2
Gene Aliases
FAM55B
UniGene
Hs.446760
Gene Chromosome Location
Chr.11: 114464276-114813702 on Build GRCh38
Transcripts
NM_152315.3 XM_011542594.2 XM_011542595.2 XM_011542596.2 XM_011542597.2 XM_011542598.2 XM_011542599.2 XM_017017207.1 XM_017017209.1 XM_017017210.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1627796 c.470-12_470-9delCTTT p.? Chr.11: 114523100-114523103 on Build GRCh38
4498483 c.520C>T p.P174S Chr.11: 114523041-114523041 on Build GRCh38
3443659 c.897G>A p.K299K Chr.11: 114523090-114523090 on Build GRCh38
1684294 c.564_566delCAA p.N189delN Chr.11: 114522995-114522997 on Build GRCh38
4498484 c.946C>T p.P316S Chr.11: 114523041-114523041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984054322 Chr.11: 114523168-114523168 on Build GRCh38
rs931615083 Chr.11: 114523156-114523156 on Build GRCh38
rs751094536 Chr.11: 114523130-114523130 on Build GRCh38
rs79538449 Chr.11: 114523035-114523035 on Build GRCh38
rs773302186 Chr.11: 114523117-114523117 on Build GRCh38
rs749330462 Chr.11: 114523071-114523071 on Build GRCh38
rs770266287 Chr.11: 114523180-114523180 on Build GRCh38
rs753307968 Chr.11: 114523079-114523080 on Build GRCh38
rs370371865 Chr.11: 114523104-114523104 on Build GRCh38
rs769814415 Chr.11: 114523111-114523111 on Build GRCh38
rs776367527 Chr.11: 114523010-114523010 on Build GRCh38
rs778296739 Chr.11: 114523066-114523066 on Build GRCh38
rs765988653 Chr.11: 114523124-114523124 on Build GRCh38
rs375564863 Chr.11: 114523143-114523143 on Build GRCh38
rs559237250 Chr.11: 114523161-114523161 on Build GRCh38
rs767121750 Chr.11: 114523141-114523141 on Build GRCh38
rs556916140 Chr.11: 114523091-114523091 on Build GRCh38
rs878966107 Chr.11: 114523137-114523137 on Build GRCh38
rs916144859 Chr.11: 114523115-114523115 on Build GRCh38
rs940097168 Chr.11: 114523181-114523181 on Build GRCh38
rs956443370 Chr.11: 114523077-114523077 on Build GRCh38
rs746107597 Chr.11: 114523092-114523092 on Build GRCh38
rs1032005129 Chr.11: 114523068-114523068 on Build GRCh38
rs749921148 Chr.11: 114523023-114523023 on Build GRCh38
rs759209567 Chr.11: 114523135-114523135 on Build GRCh38
rs77292976 Chr.11: 114523110-114523110 on Build GRCh38
rs761806769 Chr.11: 114523011-114523011 on Build GRCh38
rs267602704 Chr.11: 114523019-114523019 on Build GRCh38
rs765146411 Chr.11: 114523013-114523013 on Build GRCh38
rs771023838 Chr.11: 114523086-114523086 on Build GRCh38
rs987874259 Chr.11: 114523080-114523080 on Build GRCh38
rs766079262 Chr.11: 114523028-114523028 on Build GRCh38
rs754780976 Chr.11: 114523030-114523030 on Build GRCh38
rs377195015 Chr.11: 114523088-114523088 on Build GRCh38
rs776474530 Chr.11: 114523109-114523109 on Build GRCh38
rs762492934 Chr.11: 114523123-114523123 on Build GRCh38
rs758013670 Chr.11: 114523025-114523025 on Build GRCh38
rs908636442 Chr.11: 114523172-114523172 on Build GRCh38
rs781614218 Chr.11: 114523041-114523041 on Build GRCh38
rs756783063 Chr.11: 114523085-114523086 on Build GRCh38
rs756618760 Chr.11: 114523048-114523048 on Build GRCh38
rs781434001 Chr.11: 114523106-114523106 on Build GRCh38

View Full Product Details