Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 104037581-104037834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDI1
Entrez Gene ID
414301
Gene Name
DNA damage inducible 1 homolog 1
Gene Aliases
-
UniGene
Hs.591941
Gene Chromosome Location
Chr.11: 104036580-104039194 on Build GRCh38
Transcripts
NM_001001711.2 NM_025208.4 NM_033135.3

Gene Symbol
PDGFD
Entrez Gene ID
80310
Gene Name
platelet derived growth factor D
Gene Aliases
IEGF, MSTP036, SCDGF-B, SCDGFB
UniGene
Hs.352298
Gene Chromosome Location
Chr.11: 103907186-104164299 on Build GRCh38
Transcripts
NM_001001711.2 NM_025208.4 NM_033135.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6131158 c.860C>A p.A287D Chr.11: 104037682-104037682 on Build GRCh38
4017387 c.901C>A p.R301S Chr.11: 104037723-104037723 on Build GRCh38
4626732 c.821A>G p.E274G Chr.11: 104037643-104037643 on Build GRCh38
6641322 c.787G>A p.A263T Chr.11: 104037609-104037609 on Build GRCh38
4559177 c.795G>A p.M265I Chr.11: 104037617-104037617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988144287 Chr.11: 104037682-104037682 on Build GRCh38
rs200326028 Chr.11: 104037709-104037709 on Build GRCh38
rs762516570 Chr.11: 104037644-104037644 on Build GRCh38
rs534825593 Chr.11: 104037748-104037748 on Build GRCh38
rs759352906 Chr.11: 104037658-104037658 on Build GRCh38
rs772454143 Chr.11: 104037707-104037707 on Build GRCh38
rs763203242 Chr.11: 104037707-104037714 on Build GRCh38
rs374656084 Chr.11: 104037718-104037718 on Build GRCh38
rs751370464 Chr.11: 104037647-104037647 on Build GRCh38
rs749546050 Chr.11: 104037608-104037608 on Build GRCh38
rs1049264496 Chr.11: 104037610-104037610 on Build GRCh38
rs116423827 Chr.11: 104037789-104037789 on Build GRCh38
rs774025548 Chr.11: 104037749-104037749 on Build GRCh38
rs932829418 Chr.11: 104037751-104037751 on Build GRCh38
rs369975321 Chr.11: 104037686-104037686 on Build GRCh38
rs756071429 Chr.11: 104037673-104037673 on Build GRCh38
rs767603455 Chr.11: 104037661-104037661 on Build GRCh38
rs201356382 Chr.11: 104037807-104037807 on Build GRCh38
rs746228101 Chr.11: 104037612-104037612 on Build GRCh38
rs376710370 Chr.11: 104037615-104037615 on Build GRCh38
rs140727572 Chr.11: 104037640-104037640 on Build GRCh38
rs557940862 Chr.11: 104037776-104037776 on Build GRCh38
rs772774679 Chr.11: 104037724-104037724 on Build GRCh38
rs772649846 Chr.11: 104037642-104037642 on Build GRCh38
rs777964314 Chr.11: 104037605-104037605 on Build GRCh38
rs981047287 Chr.11: 104037645-104037645 on Build GRCh38
rs766051930 Chr.11: 104037646-104037646 on Build GRCh38
rs769083816 Chr.11: 104037723-104037723 on Build GRCh38
rs114046986 Chr.11: 104037609-104037609 on Build GRCh38
rs944100874 Chr.11: 104037721-104037721 on Build GRCh38
rs958218083 Chr.11: 104037672-104037672 on Build GRCh38
rs753260858 Chr.11: 104037722-104037722 on Build GRCh38
rs778016206 Chr.11: 104037676-104037676 on Build GRCh38
rs779169739 Chr.11: 104037696-104037696 on Build GRCh38
rs752716076 Chr.11: 104037671-104037671 on Build GRCh38
rs762511800 Chr.11: 104037743-104037743 on Build GRCh38
rs138846956 Chr.11: 104037803-104037803 on Build GRCh38
rs753892962 Chr.11: 104037680-104037680 on Build GRCh38
rs755126475 Chr.11: 104037711-104037711 on Build GRCh38
rs142985363 Chr.11: 104037697-104037697 on Build GRCh38
rs115891405 Chr.11: 104037626-104037626 on Build GRCh38
rs761129966 Chr.11: 104037628-104037628 on Build GRCh38
rs115018397 Chr.11: 104037784-104037784 on Build GRCh38

View Full Product Details