Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 8079819-8080088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF1E1
Entrez Gene ID
9521
Gene Name
eukaryotic translation elongation factor 1 epsilon 1
Gene Aliases
AIMP3, P18
UniGene
Hs.602353
Gene Chromosome Location
Chr.6: 8073360-8102595 on Build GRCh38
Transcripts
NM_001135650.1 NM_004280.4 NR_037618.1

Gene Symbol
EEF1E1-BLOC1S5
Entrez Gene ID
100526837
Gene Name
EEF1E1-BLOC1S5 readthrough (NMD candidate)
Gene Aliases
EEF1E1-MUTED
UniGene
Hs.719272
Gene Chromosome Location
Chr.6: 8013567-8102595 on Build GRCh38
Transcripts
NM_001135650.1 NM_004280.4 NR_037618.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5743112 c.484G>A p.V162I Chr.6: 8079931-8079931 on Build GRCh38
1496423 c.458C>T p.P153L Chr.6: 8079957-8079957 on Build GRCh38
7094553 c.411G>T p.E137D Chr.6: 8080004-8080004 on Build GRCh38
7210354 c.485T>C p.V162A Chr.6: 8079930-8079930 on Build GRCh38
1621967 c.426G>T p.V142V Chr.6: 8079989-8079989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192943830 Chr.6: 8079933-8079933 on Build GRCh38
rs755356428 Chr.6: 8079862-8079862 on Build GRCh38
rs756284094 Chr.6: 8079881-8079881 on Build GRCh38
rs1036092847 Chr.6: 8079865-8079865 on Build GRCh38
rs763791139 Chr.6: 8080008-8080008 on Build GRCh38
rs372034787 Chr.6: 8080042-8080042 on Build GRCh38
rs767990459 Chr.6: 8080018-8080018 on Build GRCh38
rs377588562 Chr.6: 8079875-8079875 on Build GRCh38
rs752106269 Chr.6: 8079998-8079998 on Build GRCh38
rs1003835058 Chr.6: 8079957-8079957 on Build GRCh38
rs369688934 Chr.6: 8079885-8079885 on Build GRCh38
rs940429890 Chr.6: 8079848-8079848 on Build GRCh38
rs541123053 Chr.6: 8079911-8079911 on Build GRCh38
rs764674265 Chr.6: 8079990-8079990 on Build GRCh38
rs752018094 Chr.6: 8079855-8079855 on Build GRCh38
rs774462812 Chr.6: 8079917-8079917 on Build GRCh38
rs115878881 Chr.6: 8079923-8079923 on Build GRCh38
rs201199603 Chr.6: 8080036-8080036 on Build GRCh38
rs113203747 Chr.6: 8079942-8079942 on Build GRCh38
rs763644860 Chr.6: 8079966-8079966 on Build GRCh38
rs780083950 Chr.6: 8079910-8079910 on Build GRCh38
rs61757371 Chr.6: 8079989-8079989 on Build GRCh38
rs754051616 Chr.6: 8080059-8080059 on Build GRCh38
rs777861335 Chr.6: 8080054-8080054 on Build GRCh38
rs781510788 Chr.6: 8079863-8079863 on Build GRCh38
rs762442565 Chr.6: 8079854-8079855 on Build GRCh38
rs772043441 Chr.6: 8079932-8079932 on Build GRCh38
rs1022393485 Chr.6: 8080052-8080052 on Build GRCh38
rs777966881 Chr.6: 8079883-8079883 on Build GRCh38
rs751199989 Chr.6: 8080018-8080022 on Build GRCh38
rs760445780 Chr.6: 8079955-8079955 on Build GRCh38
rs749341488 Chr.6: 8079892-8079892 on Build GRCh38
rs148223233 Chr.6: 8079908-8079908 on Build GRCh38
rs755444482 Chr.6: 8079999-8079999 on Build GRCh38
rs776293830 Chr.6: 8079984-8079984 on Build GRCh38

View Full Product Details