Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 50285813-50286086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABPB1
Entrez Gene ID
2553
Gene Name
GA binding protein transcription factor beta subunit 1
Gene Aliases
BABPB2, E4TF1, E4TF1-47, E4TF1-53, E4TF1B, GABPB, GABPB-1, GABPB2, NRF2B1, NRF2B2
UniGene
Hs.654350
Gene Chromosome Location
Chr.15: 50277192-50355429 on Build GRCh38
Transcripts
NM_001320910.1 NM_001320915.1 NM_002041.4 NM_005254.5 NM_016654.4 NM_016655.4 NM_181427.3 XM_005254274.3 XM_011521426.2 XM_017022053.1 XM_017022054.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4652604 c.1066C>T p.R356C Chr.15: 50286037-50286037 on Build GRCh38
962542 c.1019A>C p.E340A Chr.15: 50286084-50286084 on Build GRCh38
5779484 c.1018G>C p.E340Q Chr.15: 50286085-50286085 on Build GRCh38
6668491 c.1037T>C p.V346A Chr.15: 50286066-50286066 on Build GRCh38
1373359 c.1057G>A p.V353M Chr.15: 50286046-50286046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544716841 Chr.15: 50285954-50285954 on Build GRCh38
rs556085324 Chr.15: 50285869-50285872 on Build GRCh38
rs776534214 Chr.15: 50286014-50286014 on Build GRCh38
rs1055140990 Chr.15: 50285916-50285916 on Build GRCh38
rs951072562 Chr.15: 50285978-50285978 on Build GRCh38
rs768000140 Chr.15: 50286049-50286049 on Build GRCh38
rs145682817 Chr.15: 50286036-50286036 on Build GRCh38
rs1002698317 Chr.15: 50285910-50285910 on Build GRCh38
rs577350954 Chr.15: 50285926-50285926 on Build GRCh38
rs566401976 Chr.15: 50285979-50285979 on Build GRCh38
rs760015878 Chr.15: 50286005-50286006 on Build GRCh38
rs906736849 Chr.15: 50285909-50285909 on Build GRCh38
rs766337574 Chr.15: 50286019-50286019 on Build GRCh38
rs759637464 Chr.15: 50286037-50286037 on Build GRCh38
rs377427375 Chr.15: 50285982-50285982 on Build GRCh38
rs11556849 Chr.15: 50286002-50286002 on Build GRCh38
rs893776400 Chr.15: 50285935-50285938 on Build GRCh38
rs756536443 Chr.15: 50286055-50286055 on Build GRCh38
rs753055094 Chr.15: 50286050-50286050 on Build GRCh38
rs1046988167 Chr.15: 50285891-50285891 on Build GRCh38
rs369855881 Chr.15: 50285980-50285980 on Build GRCh38
rs966357698 Chr.15: 50285920-50285920 on Build GRCh38
rs555717416 Chr.15: 50285844-50285844 on Build GRCh38
rs774757664 Chr.15: 50286006-50286006 on Build GRCh38
rs1019594532 Chr.15: 50285865-50285865 on Build GRCh38
rs200123207 Chr.15: 50285971-50285971 on Build GRCh38
rs771919971 Chr.15: 50285973-50285973 on Build GRCh38
rs768519695 Chr.15: 50285992-50285992 on Build GRCh38
rs763788722 Chr.15: 50285876-50285876 on Build GRCh38

View Full Product Details