Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129040474-129040747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIGA2
Entrez Gene ID
84895
Gene Name
mitoguardin 2
Gene Aliases
C9orf54, FAM73B
UniGene
Hs.632693
Gene Chromosome Location
Chr.9: 129036605-129072082 on Build GRCh38
Transcripts
NM_032809.2 XM_005252282.1 XM_011519117.1 XM_011519118.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1105805 c.66C>A p.I22I Chr.9: 129040660-129040660 on Build GRCh38
6660143 c.26C>T p.T9M Chr.9: 129040620-129040620 on Build GRCh38
7448999 c.16G>A p.A6T Chr.9: 129040610-129040610 on Build GRCh38
4507899 c.75C>T p.F25F Chr.9: 129040669-129040669 on Build GRCh38
6895224 c.84G>A p.T28T Chr.9: 129040678-129040678 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571479593 Chr.9: 129040521-129040521 on Build GRCh38
rs746244034 Chr.9: 129040663-129040663 on Build GRCh38
rs1015714011 Chr.9: 129040523-129040523 on Build GRCh38
rs778350989 Chr.9: 129040560-129040560 on Build GRCh38
rs770666548 Chr.9: 129040702-129040702 on Build GRCh38
rs754059326 Chr.9: 129040604-129040604 on Build GRCh38
rs1022955967 Chr.9: 129040612-129040612 on Build GRCh38
rs931469072 Chr.9: 129040526-129040526 on Build GRCh38
rs748642454 Chr.9: 129040557-129040557 on Build GRCh38
rs760368945 Chr.9: 129040595-129040595 on Build GRCh38
rs759333037 Chr.9: 129040700-129040700 on Build GRCh38
rs760736796 Chr.9: 129040677-129040677 on Build GRCh38
rs764299874 Chr.9: 129040723-129040723 on Build GRCh38
rs772995314 Chr.9: 129040676-129040676 on Build GRCh38
rs747559904 Chr.9: 129040672-129040672 on Build GRCh38
rs905106170 Chr.9: 129040714-129040714 on Build GRCh38
rs767315928 Chr.9: 129040585-129040585 on Build GRCh38
rs757690925 Chr.9: 129040647-129040647 on Build GRCh38
rs762909221 Chr.9: 129040719-129040719 on Build GRCh38
rs1023976473 Chr.9: 129040681-129040681 on Build GRCh38
rs184102865 Chr.9: 129040599-129040599 on Build GRCh38
rs776379213 Chr.9: 129040700-129040700 on Build GRCh38
rs549699802 Chr.9: 129040620-129040620 on Build GRCh38
rs367611771 Chr.9: 129040559-129040559 on Build GRCh38
rs768353759 Chr.9: 129040664-129040664 on Build GRCh38
rs754904230 Chr.9: 129040627-129040627 on Build GRCh38
rs766206171 Chr.9: 129040600-129040600 on Build GRCh38
rs944027804 Chr.9: 129040510-129040510 on Build GRCh38
rs771642526 Chr.9: 129040498-129040498 on Build GRCh38
rs551560383 Chr.9: 129040520-129040520 on Build GRCh38
rs774174456 Chr.9: 129040671-129040671 on Build GRCh38
rs754112156 Chr.9: 129040547-129040547 on Build GRCh38
rs747589640 Chr.9: 129040625-129040625 on Build GRCh38
rs776352320 Chr.9: 129040680-129040680 on Build GRCh38
rs746626918 Chr.9: 129040648-129040648 on Build GRCh38
rs749015669 Chr.9: 129040575-129040575 on Build GRCh38
rs778117776 Chr.9: 129040621-129040621 on Build GRCh38
rs193049423 Chr.9: 129040499-129040499 on Build GRCh38
rs531639563 Chr.9: 129040507-129040507 on Build GRCh38
rs768264076 Chr.9: 129040581-129040581 on Build GRCh38
rs765405535 Chr.9: 129040709-129040709 on Build GRCh38
rs761388053 Chr.9: 129040584-129040584 on Build GRCh38
rs753011054 Chr.9: 129040710-129040710 on Build GRCh38
rs547099120 Chr.9: 129040673-129040673 on Build GRCh38
rs1048588017 Chr.9: 129040539-129040539 on Build GRCh38
rs773226774 Chr.9: 129040591-129040591 on Build GRCh38
rs765458799 Chr.9: 129040615-129040615 on Build GRCh38
rs775251905 Chr.9: 129040566-129040566 on Build GRCh38
rs771374590 Chr.9: 129040563-129040563 on Build GRCh38
rs752654829 Chr.9: 129040618-129040618 on Build GRCh38
rs773937481 Chr.9: 129040582-129040582 on Build GRCh38
rs755023414 Chr.9: 129040553-129040553 on Build GRCh38
rs146566671 Chr.9: 129040678-129040678 on Build GRCh38
rs755252265 Chr.9: 129040613-129040613 on Build GRCh38
rs778334502 Chr.9: 129040517-129040517 on Build GRCh38
rs779035053 Chr.9: 129040556-129040556 on Build GRCh38

View Full Product Details