Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73293047-73293282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGR4
Entrez Gene ID
1961
Gene Name
early growth response 4
Gene Aliases
NGFI-C, NGFIC, PAT133
UniGene
Hs.3052
Gene Chromosome Location
Chr.2: 73290929-73293701 on Build GRCh38
Transcripts
NM_001965.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6427602 c.115G>T p.A39S Chr.2: 73293203-73293203 on Build GRCh38
6427601 c.424G>T p.A142S Chr.2: 73293203-73293203 on Build GRCh38
1205066 c.425C>T p.A142V Chr.2: 73293202-73293202 on Build GRCh38
6455428 c.120C>A p.A40A Chr.2: 73293198-73293198 on Build GRCh38
1409391 c.414G>T p.R138S Chr.2: 73293213-73293213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748358260 Chr.2: 73293137-73293137 on Build GRCh38
rs555396652 Chr.2: 73293212-73293212 on Build GRCh38
rs978912217 Chr.2: 73293216-73293216 on Build GRCh38
rs773249919 Chr.2: 73293152-73293152 on Build GRCh38
rs969398077 Chr.2: 73293119-73293119 on Build GRCh38
rs775445367 Chr.2: 73293173-73293173 on Build GRCh38
rs532431372 Chr.2: 73293101-73293101 on Build GRCh38
rs571777946 Chr.2: 73293077-73293077 on Build GRCh38
rs145377731 Chr.2: 73293189-73293189 on Build GRCh38
rs557977459 Chr.2: 73293110-73293110 on Build GRCh38
rs897630457 Chr.2: 73293109-73293109 on Build GRCh38
rs972610149 Chr.2: 73293154-73293154 on Build GRCh38
rs779302657 Chr.2: 73293238-73293238 on Build GRCh38
rs550150527 Chr.2: 73293065-73293065 on Build GRCh38
rs550654844 Chr.2: 73293127-73293127 on Build GRCh38
rs960828001 Chr.2: 73293091-73293091 on Build GRCh38
rs747145988 Chr.2: 73293163-73293163 on Build GRCh38
rs756320734 Chr.2: 73293231-73293232 on Build GRCh38
rs533954745 Chr.2: 73293187-73293187 on Build GRCh38
rs779166677 Chr.2: 73293132-73293132 on Build GRCh38
rs762823394 Chr.2: 73293186-73293186 on Build GRCh38
rs958441651 Chr.2: 73293232-73293232 on Build GRCh38
rs377552654 Chr.2: 73293171-73293171 on Build GRCh38
rs925727029 Chr.2: 73293218-73293218 on Build GRCh38
rs865991064 Chr.2: 73293230-73293230 on Build GRCh38
rs377352409 Chr.2: 73293240-73293240 on Build GRCh38
rs749947757 Chr.2: 73293200-73293200 on Build GRCh38
rs1043972656 Chr.2: 73293227-73293227 on Build GRCh38
rs375301544 Chr.2: 73293188-73293188 on Build GRCh38
rs370512012 Chr.2: 73293248-73293248 on Build GRCh38
rs1018273308 Chr.2: 73293158-73293158 on Build GRCh38
rs180920424 Chr.2: 73293100-73293100 on Build GRCh38
rs1027049024 Chr.2: 73293120-73293120 on Build GRCh38
rs151267217 Chr.2: 73293226-73293226 on Build GRCh38
rs373746151 Chr.2: 73293147-73293147 on Build GRCh38
rs748351654 Chr.2: 73293229-73293231 on Build GRCh38
rs747250056 Chr.2: 73293251-73293251 on Build GRCh38
rs765998769 Chr.2: 73293227-73293227 on Build GRCh38
rs755467704 Chr.2: 73293233-73293233 on Build GRCh38
rs1031482551 Chr.2: 73293084-73293084 on Build GRCh38
rs754319562 Chr.2: 73293229-73293229 on Build GRCh38
rs999658445 Chr.2: 73293231-73293231 on Build GRCh38
rs140526933 Chr.2: 73293255-73293255 on Build GRCh38
rs772318013 Chr.2: 73293139-73293139 on Build GRCh38

View Full Product Details