Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 38929155-38929424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM114A1
Entrez Gene ID
92689
Gene Name
family with sequence similarity 114 member A1
Gene Aliases
Noxp20
UniGene
Hs.476517
Gene Chromosome Location
Chr.4: 38867733-38945744 on Build GRCh38
Transcripts
NM_138389.2 NR_033290.1 XM_005262672.2 XM_006714033.2 XM_017008838.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5624932 c.1117C>G p.L373V Chr.4: 38929289-38929289 on Build GRCh38
6026967 c.1161+3C>T p.? Chr.4: 38929336-38929336 on Build GRCh38
1284805 c.1131T>A p.H377Q Chr.4: 38929303-38929303 on Build GRCh38
5060338 c.1104G>T p.M368I Chr.4: 38929276-38929276 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370970875 Chr.4: 38929378-38929378 on Build GRCh38
rs748668113 Chr.4: 38929287-38929287 on Build GRCh38
rs376820553 Chr.4: 38929227-38929227 on Build GRCh38
rs774762685 Chr.4: 38929182-38929182 on Build GRCh38
rs66855132 Chr.4: 38929374-38929374 on Build GRCh38
rs973066193 Chr.4: 38929345-38929345 on Build GRCh38
rs199734440 Chr.4: 38929271-38929271 on Build GRCh38
rs779577673 Chr.4: 38929372-38929372 on Build GRCh38
rs921228573 Chr.4: 38929277-38929278 on Build GRCh38
rs777569993 Chr.4: 38929308-38929308 on Build GRCh38
rs763559017 Chr.4: 38929368-38929369 on Build GRCh38
rs762359801 Chr.4: 38929188-38929188 on Build GRCh38
rs776747191 Chr.4: 38929309-38929309 on Build GRCh38
rs754064027 Chr.4: 38929218-38929218 on Build GRCh38
rs749672493 Chr.4: 38929177-38929177 on Build GRCh38
rs372319298 Chr.4: 38929214-38929214 on Build GRCh38
rs750337870 Chr.4: 38929356-38929356 on Build GRCh38
rs759153786 Chr.4: 38929320-38929327 on Build GRCh38
rs58035134 Chr.4: 38929374-38929374 on Build GRCh38
rs767500824 Chr.4: 38929343-38929343 on Build GRCh38
rs750757129 Chr.4: 38929365-38929365 on Build GRCh38
rs963254753 Chr.4: 38929311-38929311 on Build GRCh38
rs755638502 Chr.4: 38929257-38929257 on Build GRCh38
rs200191699 Chr.4: 38929219-38929219 on Build GRCh38
rs745978479 Chr.4: 38929375-38929375 on Build GRCh38
rs770139256 Chr.4: 38929388-38929388 on Build GRCh38
rs778703287 Chr.4: 38929376-38929376 on Build GRCh38
rs577366890 Chr.4: 38929342-38929342 on Build GRCh38
rs759752625 Chr.4: 38929325-38929325 on Build GRCh38
rs3188469 Chr.4: 38929300-38929300 on Build GRCh38
rs772288907 Chr.4: 38929210-38929210 on Build GRCh38
rs780689448 Chr.4: 38929250-38929250 on Build GRCh38
rs750005970 Chr.4: 38929256-38929256 on Build GRCh38
rs751522847 Chr.4: 38929226-38929226 on Build GRCh38
rs35369040 Chr.4: 38929364-38929365 on Build GRCh38
rs557486959 Chr.4: 38929302-38929302 on Build GRCh38
rs949569337 Chr.4: 38929211-38929211 on Build GRCh38
rs368326921 Chr.4: 38929212-38929212 on Build GRCh38
rs190866449 Chr.4: 38929362-38929362 on Build GRCh38
rs896621445 Chr.4: 38929196-38929196 on Build GRCh38
rs376563547 Chr.4: 38929329-38929329 on Build GRCh38
rs397969757 Chr.4: 38929373-38929374 on Build GRCh38
rs769045161 Chr.4: 38929181-38929181 on Build GRCh38
rs376022629 Chr.4: 38929374-38929375 on Build GRCh38
rs748580512 Chr.4: 38929381-38929381 on Build GRCh38
rs752380827 Chr.4: 38929215-38929215 on Build GRCh38
rs149149187 Chr.4: 38929269-38929269 on Build GRCh38
rs77257789 Chr.4: 38929222-38929222 on Build GRCh38
rs779361483 Chr.4: 38929280-38929280 on Build GRCh38
rs372821613 Chr.4: 38929327-38929327 on Build GRCh38
rs2306923 Chr.4: 38929272-38929272 on Build GRCh38
rs772773424 Chr.4: 38929296-38929296 on Build GRCh38

View Full Product Details