Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63589923-63590196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMEB2
Entrez Gene ID
26205
Gene Name
glucocorticoid modulatory element binding protein 2
Gene Aliases
GMEB-2, P79PIF, PIF79
UniGene
Hs.473286
Gene Chromosome Location
Chr.20: 63587594-63627101 on Build GRCh38
Transcripts
NM_012384.4 XM_005260202.4 XM_006723776.2 XM_011528779.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7130058 c.1534G>A p.E512K Chr.20: 63590148-63590148 on Build GRCh38
7130057 c.1573G>C p.E525Q Chr.20: 63590109-63590109 on Build GRCh38
6004634 c.1564G>A p.A522T Chr.20: 63590118-63590118 on Build GRCh38
4100444 c.1540C>T p.H514Y Chr.20: 63590142-63590142 on Build GRCh38
478395 c.1513G>A p.A505T Chr.20: 63590169-63590169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776758045 Chr.20: 63590169-63590169 on Build GRCh38
rs770598865 Chr.20: 63590070-63590070 on Build GRCh38
rs973542159 Chr.20: 63590100-63590100 on Build GRCh38
rs771330766 Chr.20: 63590155-63590155 on Build GRCh38
rs774828421 Chr.20: 63589985-63589985 on Build GRCh38
rs1004482861 Chr.20: 63590131-63590131 on Build GRCh38
rs762714185 Chr.20: 63589998-63589998 on Build GRCh38
rs192220884 Chr.20: 63590035-63590035 on Build GRCh38
rs891633344 Chr.20: 63590034-63590034 on Build GRCh38
rs771556140 Chr.20: 63590158-63590158 on Build GRCh38
rs143150017 Chr.20: 63590076-63590076 on Build GRCh38
rs112328838 Chr.20: 63590052-63590052 on Build GRCh38
rs760652797 Chr.20: 63590163-63590163 on Build GRCh38
rs376285096 Chr.20: 63590075-63590075 on Build GRCh38
rs748889830 Chr.20: 63590069-63590069 on Build GRCh38
rs781716427 Chr.20: 63590054-63590054 on Build GRCh38
rs145516314 Chr.20: 63590138-63590138 on Build GRCh38
rs748595394 Chr.20: 63590144-63590144 on Build GRCh38
rs2297427 Chr.20: 63590011-63590011 on Build GRCh38
rs757159681 Chr.20: 63590097-63590097 on Build GRCh38
rs920825934 Chr.20: 63589991-63589991 on Build GRCh38
rs769411088 Chr.20: 63589977-63589977 on Build GRCh38
rs201239578 Chr.20: 63590050-63590050 on Build GRCh38
rs376020745 Chr.20: 63590062-63590062 on Build GRCh38
rs748669759 Chr.20: 63590056-63590056 on Build GRCh38
rs140964265 Chr.20: 63590175-63590175 on Build GRCh38
rs980259466 Chr.20: 63590027-63590027 on Build GRCh38
rs556258139 Chr.20: 63590022-63590022 on Build GRCh38
rs927059299 Chr.20: 63589999-63589999 on Build GRCh38
rs764092751 Chr.20: 63590165-63590165 on Build GRCh38
rs886119763 Chr.20: 63590122-63590122 on Build GRCh38
rs544962738 Chr.20: 63590036-63590036 on Build GRCh38
rs774036963 Chr.20: 63590157-63590157 on Build GRCh38
rs900385146 Chr.20: 63589983-63589983 on Build GRCh38
rs551655648 Chr.20: 63590160-63590160 on Build GRCh38
rs567384252 Chr.20: 63589964-63589964 on Build GRCh38
rs759060075 Chr.20: 63590073-63590073 on Build GRCh38
rs770077487 Chr.20: 63590061-63590061 on Build GRCh38
rs751787794 Chr.20: 63590113-63590113 on Build GRCh38
rs766602123 Chr.20: 63590101-63590101 on Build GRCh38
rs908031665 Chr.20: 63589965-63589966 on Build GRCh38
rs997838397 Chr.20: 63589989-63589989 on Build GRCh38
rs753718087 Chr.20: 63590096-63590096 on Build GRCh38
rs373395219 Chr.20: 63590086-63590086 on Build GRCh38
rs779958855 Chr.20: 63590038-63590038 on Build GRCh38
rs756634351 Chr.20: 63590146-63590146 on Build GRCh38
rs778068236 Chr.20: 63590149-63590149 on Build GRCh38
rs140213772 Chr.20: 63590137-63590137 on Build GRCh38
rs993061384 Chr.20: 63590049-63590049 on Build GRCh38
rs954991698 Chr.20: 63590066-63590066 on Build GRCh38
rs755080232 Chr.20: 63590124-63590124 on Build GRCh38
rs763935412 Chr.20: 63590087-63590087 on Build GRCh38
rs963193624 Chr.20: 63590141-63590141 on Build GRCh38
rs773911472 Chr.20: 63590071-63590071 on Build GRCh38
rs1029994946 Chr.20: 63589995-63589995 on Build GRCh38
rs1015938976 Chr.20: 63590021-63590021 on Build GRCh38
rs201863145 Chr.20: 63590154-63590154 on Build GRCh38

View Full Product Details