Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37373880-37374121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELFN2
Entrez Gene ID
114794
Gene Name
extracellular leucine rich repeat and fibronectin type III domain containing 2
Gene Aliases
LRRC62, PPP1R29
UniGene
Hs.344170
Gene Chromosome Location
Chr.22: 37340644-37427470 on Build GRCh38
Transcripts
NM_052906.4 NR_110512.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680756 c.1517G>A p.R506H Chr.22: 37374018-37374018 on Build GRCh38
7189900 c.1498G>A p.G500S Chr.22: 37374037-37374037 on Build GRCh38
4680755 c.1565C>T p.P522L Chr.22: 37373970-37373970 on Build GRCh38
5948109 c.1531G>A p.G511R Chr.22: 37374004-37374004 on Build GRCh38
726382 c.1552G>A p.E518K Chr.22: 37373983-37373983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753998297 Chr.22: 37374092-37374092 on Build GRCh38
rs8142615 Chr.22: 37374026-37374026 on Build GRCh38
rs773698358 Chr.22: 37374081-37374081 on Build GRCh38
rs746549719 Chr.22: 37373988-37373988 on Build GRCh38
rs753856853 Chr.22: 37374021-37374021 on Build GRCh38
rs772799053 Chr.22: 37373984-37373984 on Build GRCh38
rs779860760 Chr.22: 37373963-37373963 on Build GRCh38
rs137984700 Chr.22: 37373939-37373939 on Build GRCh38
rs766538809 Chr.22: 37374020-37374020 on Build GRCh38
rs201073943 Chr.22: 37374086-37374086 on Build GRCh38
rs764480223 Chr.22: 37374005-37374005 on Build GRCh38
rs772746026 Chr.22: 37373905-37373905 on Build GRCh38
rs751868133 Chr.22: 37373948-37373948 on Build GRCh38
rs756161403 Chr.22: 37374015-37374015 on Build GRCh38
rs376827195 Chr.22: 37373970-37373970 on Build GRCh38
rs764581745 Chr.22: 37373946-37373946 on Build GRCh38
rs566547957 Chr.22: 37373956-37373956 on Build GRCh38
rs746725384 Chr.22: 37374052-37374052 on Build GRCh38
rs376193256 Chr.22: 37374072-37374072 on Build GRCh38
rs749190458 Chr.22: 37373969-37373969 on Build GRCh38
rs56845582 Chr.22: 37373962-37373962 on Build GRCh38
rs149473582 Chr.22: 37374004-37374004 on Build GRCh38
rs560850924 Chr.22: 37373979-37373979 on Build GRCh38
rs542011949 Chr.22: 37373957-37373957 on Build GRCh38
rs776247026 Chr.22: 37374064-37374064 on Build GRCh38
rs368368952 Chr.22: 37373981-37373981 on Build GRCh38
rs754982133 Chr.22: 37374022-37374022 on Build GRCh38
rs770420117 Chr.22: 37373998-37373998 on Build GRCh38
rs776311867 Chr.22: 37374063-37374063 on Build GRCh38
rs764482250 Chr.22: 37374060-37374060 on Build GRCh38
rs146010699 Chr.22: 37374008-37374008 on Build GRCh38
rs1038197033 Chr.22: 37373951-37373951 on Build GRCh38
rs758144404 Chr.22: 37374027-37374027 on Build GRCh38
rs143903281 Chr.22: 37374007-37374007 on Build GRCh38
rs745351154 Chr.22: 37374067-37374067 on Build GRCh38
rs372178445 Chr.22: 37373945-37373945 on Build GRCh38
rs201585205 Chr.22: 37374011-37374011 on Build GRCh38
rs199741364 Chr.22: 37373983-37373983 on Build GRCh38
rs765681278 Chr.22: 37373943-37373943 on Build GRCh38
rs777403678 Chr.22: 37374051-37374051 on Build GRCh38
rs201726267 Chr.22: 37373999-37373999 on Build GRCh38
rs61743097 Chr.22: 37374068-37374068 on Build GRCh38
rs762273969 Chr.22: 37374077-37374077 on Build GRCh38
rs757596776 Chr.22: 37373952-37373952 on Build GRCh38
rs140163106 Chr.22: 37374085-37374085 on Build GRCh38

View Full Product Details