Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15289364-15289617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHAD1
Entrez Gene ID
114827
Gene Name
forkhead associated phosphopeptide binding domain 1
Gene Aliases
-
UniGene
Hs.659997
Gene Chromosome Location
Chr.1: 15236559-15400283 on Build GRCh38
Transcripts
NM_052929.1 XM_011540575.2 XM_011540576.2 XM_011540577.2 XM_011540580.2 XM_011540581.2 XM_011540582.2 XM_011540584.2 XM_011540588.2 XM_011540589.2 XM_011540590.2 XM_011540592.2 XM_011540593.2 XM_011540595.2 XM_011540597.2 XM_011540598.2 XM_017000198.1 XM_017000199.1 XM_017000200.1 XM_017000201.1 XM_017000202.1 XM_017000203.1 XM_017000204.1 XM_017000205.1 XM_017000206.1 XM_017000207.1 XM_017000208.1 XM_017000209.1 XM_017000210.1 XM_017000211.1 XM_017000212.1 XM_017000213.1 XM_017000214.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3979891 c.336G>A p.W112* Chr.1: 15289434-15289434 on Build GRCh38
4491688 c.384C>T p.P128P Chr.1: 15289482-15289482 on Build GRCh38
4334857 c.380delC p.P129fs*39 Chr.1: 15289478-15289478 on Build GRCh38
4453944 c.373A>C p.N125H Chr.1: 15289471-15289471 on Build GRCh38
4142810 c.383C>A p.P128H Chr.1: 15289481-15289481 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965275161 Chr.1: 15289393-15289393 on Build GRCh38
rs368473934 Chr.1: 15289479-15289479 on Build GRCh38
rs920877849 Chr.1: 15289478-15289478 on Build GRCh38
rs542248223 Chr.1: 15289410-15289410 on Build GRCh38
rs755826698 Chr.1: 15289509-15289509 on Build GRCh38
rs543339451 Chr.1: 15289522-15289522 on Build GRCh38
rs756645134 Chr.1: 15289462-15289462 on Build GRCh38
rs200337988 Chr.1: 15289494-15289494 on Build GRCh38
rs945291224 Chr.1: 15289595-15289595 on Build GRCh38
rs754525732 Chr.1: 15289566-15289566 on Build GRCh38
rs561909148 Chr.1: 15289484-15289484 on Build GRCh38
rs927808997 Chr.1: 15289596-15289596 on Build GRCh38
rs753222513 Chr.1: 15289457-15289457 on Build GRCh38
rs781623345 Chr.1: 15289592-15289592 on Build GRCh38
rs778250154 Chr.1: 15289577-15289577 on Build GRCh38
rs563528811 Chr.1: 15289526-15289526 on Build GRCh38
rs763173102 Chr.1: 15289438-15289438 on Build GRCh38
rs761683608 Chr.1: 15289488-15289488 on Build GRCh38
rs753326995 Chr.1: 15289559-15289559 on Build GRCh38
rs749610562 Chr.1: 15289500-15289500 on Build GRCh38
rs756234754 Chr.1: 15289477-15289478 on Build GRCh38
rs747573827 Chr.1: 15289578-15289578 on Build GRCh38
rs532174986 Chr.1: 15289527-15289527 on Build GRCh38
rs574694413 Chr.1: 15289485-15289485 on Build GRCh38
rs973609998 Chr.1: 15289448-15289448 on Build GRCh38
rs200038906 Chr.1: 15289591-15289591 on Build GRCh38
rs764090587 Chr.1: 15289454-15289454 on Build GRCh38
rs372898600 Chr.1: 15289414-15289414 on Build GRCh38

View Full Product Details