Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 69251226-69251477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF6
Entrez Gene ID
11052
Gene Name
cleavage and polyadenylation specific factor 6
Gene Aliases
CFIM, CFIM68, HPBRII-4, HPBRII-7
UniGene
Hs.369606
Gene Chromosome Location
Chr.12: 69239537-69274358 on Build GRCh38
Transcripts
NM_001300947.1 NM_007007.2 XM_005268588.2 XM_005268590.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5613602 c.187G>T p.D63Y Chr.12: 69251255-69251255 on Build GRCh38
3399034 c.193G>C p.G65R Chr.12: 69251261-69251261 on Build GRCh38
1363889 c.214G>A p.V72I Chr.12: 69251282-69251282 on Build GRCh38
3704386 c.201A>G p.G67G Chr.12: 69251269-69251269 on Build GRCh38
7317746 c.239G>A p.R80K Chr.12: 69251307-69251307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2231692 Chr.12: 69251449-69251449 on Build GRCh38
rs571322908 Chr.12: 69251349-69251349 on Build GRCh38
rs900625487 Chr.12: 69251330-69251330 on Build GRCh38
rs201287830 Chr.12: 69251279-69251279 on Build GRCh38
rs539174741 Chr.12: 69251450-69251452 on Build GRCh38
rs200812392 Chr.12: 69251382-69251382 on Build GRCh38
rs1003372534 Chr.12: 69251417-69251417 on Build GRCh38
rs751772212 Chr.12: 69251285-69251285 on Build GRCh38
rs10784773 Chr.12: 69251455-69251455 on Build GRCh38
rs1049065156 Chr.12: 69251369-69251369 on Build GRCh38
rs769018336 Chr.12: 69251348-69251348 on Build GRCh38
rs370987967 Chr.12: 69251261-69251261 on Build GRCh38
rs748967800 Chr.12: 69251354-69251354 on Build GRCh38
rs377589979 Chr.12: 69251395-69251395 on Build GRCh38
rs375695206 Chr.12: 69251346-69251346 on Build GRCh38
rs930712444 Chr.12: 69251424-69251424 on Build GRCh38
rs372491033 Chr.12: 69251376-69251376 on Build GRCh38
rs141570992 Chr.12: 69251296-69251296 on Build GRCh38
rs78419067 Chr.12: 69251284-69251284 on Build GRCh38
rs534673554 Chr.12: 69251293-69251293 on Build GRCh38
rs780654477 Chr.12: 69251314-69251314 on Build GRCh38
rs751911807 Chr.12: 69251394-69251394 on Build GRCh38
rs555168127 Chr.12: 69251403-69251403 on Build GRCh38
rs573808651 Chr.12: 69251441-69251441 on Build GRCh38
rs962046469 Chr.12: 69251384-69251384 on Build GRCh38
rs370014669 Chr.12: 69251373-69251373 on Build GRCh38
rs541014666 Chr.12: 69251445-69251445 on Build GRCh38
rs543644513 Chr.12: 69251401-69251401 on Build GRCh38
rs961261366 Chr.12: 69251412-69251412 on Build GRCh38
rs756618819 Chr.12: 69251453-69251453 on Build GRCh38
rs911073900 Chr.12: 69251451-69251451 on Build GRCh38
rs539324402 Chr.12: 69251358-69251358 on Build GRCh38

View Full Product Details